Skip to Content
Merck
All Photos(1)

Key Documents

W318101

Sigma-Aldrich

2-Methoxycinnamaldehyde

greener alternative

natural, 98%, FG

Synonym(s):

o-Methoxycinnamaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH=CHCHO
CAS Number:
Molecular Weight:
162.19
FEMA Number:
3181
EC Number:
Council of Europe no.:
571
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.048
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

160-161 °C/12 mmHg (lit.)

mp

44.0-49.0 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cinnamon; woody; spicy; sweet

SMILES string

[H]C(=O)C=Cc1ccccc1OC

InChI

1S/C10H10O2/c1-12-10-7-3-2-5-9(10)6-4-8-11/h2-8H,1H3/b6-4+

InChI key

KKVZAVRSVHUSPL-GQCTYLIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Network pharmacology combined with molecular docking and experimental validation to explore the potential mechanism of Cinnamomi ramulus against ankylosing spondylitis.: This study investigates the anti-inflammatory potential of 2-Methoxycinnamaldehyde in Cinnamomi ramulus. Its application extends to novel therapeutic approaches for treating ankylosing spondylitis, demonstrating significant implications for medicinal chemistry and pharmacology (Wei et al., 2023).

  • Ramulus Cinnamomi essential oil exerts an anti-inflammatory effect on RAW264.7 cells through N-acylethanolamine acid amidase inhibition.: The study elaborates on the anti-inflammatory activities of 2-Methoxycinnamaldehyde, offering a biochemical pathway that could be exploited in anti-inflammatory drug design (Jia et al., 2023).
  • Metabolomics-Driven Exploration of the Antibacterial Activity and Mechanism of 2-Methoxycinnamaldehyde.: This article offers insights into the antibacterial properties of 2-Methoxycinnamaldehyde, using metabolomics to explore its mechanism of action, significant for developments in antimicrobial treatments (Qian et al., 2022).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-you Guo et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 31(13), 1087-1090 (2006-10-20)
To observe the effect of 2-methoxycinnamaldehyde (isolated from fraction A of Guizhi Tang) on activity of COX and PGE2 release in rat cerebral microvascular endothelial cells (rCMEC) stimulated by IL-1. rCMEC were cultured, and identified by immunohistochemistry for von Willebrand
R Giordani et al.
Phytotherapy research : PTR, 20(1), 58-61 (2006-01-07)
The antifungal activity of the essential oil from Cinnamomum cassia, alone or combined with amphotericin B, a drug widely used for most indications despite side-effects was investigated. The composition of the oil was analysed by GC/MS and characterized by its
Alavala Matta Reddy et al.
Planta medica, 70(9), 823-827 (2004-10-27)
Nuclear factor (NF)-cB is a transcription factor regulating the expression of inflammatory and immune genes. In the present study, an extract from stem bark of Cinnamomum cassia Blume(Lauraceae) was discovered to have an inhibitory effect on LPS-induced NF-KB transcriptional activity
Guang-Ping Lv et al.
Journal of separation science, 33(15), 2341-2348 (2010-06-24)
A pressurized liquid extraction and GC-MS method was developed for simultaneous quantitative determination of the seven components, including cinnamaldehyde, copaene, cinnamic acid, coumarin, 2-methoxycinnamaldehyde, 2-methoxycinnamic acid and safrole in Cinnamomum cassia. The results showed that methanol and ethanol was not
Jian-You Guo et al.
Biological & pharmaceutical bulletin, 29(11), 2214-2221 (2006-11-02)
Prostaglandin E2 (PGE2) works as a common final mediator of the febrile. Guizhi-Tang, one of the most famous traditional Chinese medical formula used to treat influenza, common cold and other pyretic conditions, was previously reported to reduce the production of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service