Skip to Content
Merck
All Photos(1)

Documents

C80687

Sigma-Aldrich

trans-Cinnamaldehyde

97%

Synonym(s):

trans-3-Phenyl-2-propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
Beilstein:
1071571
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.6 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.622 (lit.)

bp

250-252 °C (lit.)

mp

−9-−4 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)\C=C\c1ccccc1

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-Cinnamaldehyde has been used:
  • To synthesize trans-cinnamaldehyde -β-cyclodextrin complex, an antimicrobial edible coating which can increase the shelf life of fresh-cut fruits.

It can be used:
  • As a precursor to synthesize functionalized furans.
  • For the synthesis of hydrocinnamyl alcohol by hydrogenation catalyzed by SBA-15-supported copper–chromium mixed oxide nanoparticles (CuCr/SBA-15).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multilayered antimicrobial edible coating and its effect on quality and shelf-life of fresh-cut pineapple (Ananas comosus).
Mantilla N, et al.
LWT--Food Science and Technology, 51(1), 37-43 (2013)
Enhancing the performance of SBA-15-supported copper catalysts by chromium addition for the chemoselective hydrogenation of trans-cinnamaldehyde.
Dragoi B, et al.
Catalysis Science & Technology, 3(9), 2319-2329 (2013)
A Facile One-Pot Synthesis of Amino Furans Using Trans-Cinnamaldehyde in the Presence of Nucleophilic Isocyanides.
Asghari S and Qandalee M
Acta Chimica Slovenica, 54(3), 341-350 (2007)
Polysaccharide-based multilayered antimicrobial edible coating enhances quality of fresh-cut papaya.
Brasil IM, et al.
LWT--Food Science and Technology, 47(1), 39-45 (2012)
Improved multilayered antimicrobial alginate-based edible coating extends the shelf life of fresh-cut watermelon (Citrullus lanatus).
Sipahi RE, et al.
LWT--Food Science and Technology, 51(1), 9-15 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service