Skip to Content
Merck
All Photos(3)

Key Documents

W278513

Sigma-Aldrich

2-Nonanone

greener alternative

natural, ≥97%, FCC, FG

Synonym(s):

Heptyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COCH3
CAS Number:
Molecular Weight:
142.24
FEMA Number:
2785
Beilstein:
1743645
EC Number:
Council of Europe no.:
154
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.020
NACRES:
NA.21
Organoleptic:
cheese; coconut; green; fruity; waxy; herbaceous
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥97%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.421 (lit.)

bp

192 °C/743 mmHg (lit.)

mp

−21 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cheese; coconut; green; fruity; waxy; herbaceous

SMILES string

CCCCCCCC(C)=O

InChI

1S/C9H18O/c1-3-4-5-6-7-8-9(2)10/h3-8H2,1-2H3

InChI key

VKCYHJWLYTUGCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural occurrence: Blue cheese, butter, cheddar cheese, coconut, fish, krill, oil of rue.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Antifungal activity of bio-active cell-free culture extracts and volatile organic compounds (VOCs) synthesised by endophytic fungal isolates of Garden Nasturtium.: This research highlights the antifungal properties of 2-Nonanone, demonstrating its potential in biological control and offering an alternative to chemical pesticides in agricultural practices (Santra et al., 2024).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The neurotoxicity of 5-nonanone: preliminary report.
M A Shifman et al.
Toxicology letters, 8(4-5), 283-288 (1981-06-01)
E R Troemel et al.
Cell, 91(2), 161-169 (1997-11-05)
Different olfactory cues elicit distinct behaviors such as attraction, avoidance, feeding, or mating. In the nematode C. elegans, these cues are sensed by a small number of olfactory neurons, each of which expresses several different odorant receptors. The type of
T D Dreesen et al.
The Biochemical journal, 203(1), 69-75 (1982-04-01)
Individual turbinals from the right and left sides of dog olfactory tissue were removed and nerve-ending-particle preparations were prepared. (Na+ + K+)-dependent ATPase activities of the individual preparations, and the effect of several odorous compounds [including (+)- and (-)-carvone] on
D A Birnby et al.
Genetics, 155(1), 85-104 (2000-05-03)
Caenorhabditis elegans daf-11 and daf-21 mutants share defects in specific chemosensory responses mediated by several classes of sensory neurons, indicating that these two genes have closely related functions in an assortment of chemosensory pathways. We report that daf-11 encodes one
J A Dunn et al.
Experimental & applied acarology, 37(1-2), 131-140 (2005-09-24)
Two types of trap, the BT Trap and PC(TM) pitfall cone trap, were evaluated against the most widely used method of sampling and sieving for their ability to detect low numbers of storage mites (Acarus siro, Tyrophagus longior and Lepidoglyphus

Articles

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

The EU’s definition of natural flavors is under review as GMM use in fermentation raises questions. Learn how regulations and consumer perceptions may shift.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service