Skip to Content
Merck
All Photos(1)

Key Documents

W272906

Sigma-Aldrich

Methyl 2-octynoate

≥99%, stabilized, FCC, FG

Synonym(s):

Methyl heptine carbonate, NSC 72098

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C≡CCO2CH3
CAS Number:
Molecular Weight:
154.21
FEMA Number:
2729
Beilstein:
1756887
EC Number:
Council of Europe no.:
481
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.158
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥99%

contains

α-tocopherol, synthetic as stabilizer

composition

contains IFRA and 1223/2009 restricted Methyl 2-octynoate

refractive index

n20/D 1.446 (lit.)

bp

217-220 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; vegetable; violet

SMILES string

CCCCCC#CC(=O)OC

InChI

1S/C9H14O2/c1-3-4-5-6-7-8-9(10)11-2/h3-6H2,1-2H3

InChI key

FRLZQXRXIKQFNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Skin Sens. 1A

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Takanami et al.
Gan no rinsho. Japan journal of cancer clinics, 30(4), 340-343 (1984-04-01)
Results of methyl heptine carbonate skin tests were compared with DNCB skin test results in 155 patients with chest diseases. Reactions to methyl heptine carbonate and DNCB were similar in 142 of 155 patients (92%). Forty-two lung cancer patients and
Jurij J Hostynek et al.
Cutaneous and ocular toxicology, 25(4), 259-271 (2006-12-13)
The fragrance material methyl heptine carbonate has been cited as a moderately frequent cause of cosmetics-related allergic contact dermatitis. A review of the literature shows that when the underlying clinical data are analyzed, there is only one published case where
[Cutaneous delayed hypersensitivity to methyl hepatine carbonate in pulmonary tuberculosis].
I Takanami
Nihon Kyobu Shikkan Gakkai zasshi, 24(10), 1066-1070 (1986-10-01)
Andre H St Amant et al.
Journal of the American Chemical Society, 141(34), 13619-13624 (2019-08-08)
Herein, we report the development of a scalable and synthetically robust building block based on norbornadiene (NBD) that can be broadly incorporated into a variety of macromolecular architectures using traditional living polymerization techniques. By taking advantage of a selective and
Allergic contact dermatitis from methyl heptine and methyl octine carbonates.
J S English et al.
Contact dermatitis, 18(3), 174-175 (1988-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service