Skip to Content
Merck
All Photos(1)

Key Documents

W267708

Sigma-Aldrich

4′-Methylacetophenone

≥95%, FCC, FG

Synonym(s):

Methyl p-tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
FEMA Number:
2677
Beilstein:
606053
EC Number:
Council of Europe no.:
156
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.022
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.533 (lit.)

bp

226 °C (lit.)

mp

22-24 °C (lit.)

density

1.004 g/mL at 20 °C
1.005 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

hawthorne; floral

SMILES string

CC(=O)c1ccc(C)cc1

InChI

1S/C9H10O/c1-7-3-5-9(6-4-7)8(2)10/h3-6H,1-2H3

InChI key

GNKZMNRKLCTJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4′-Methylacetophenone occurs naturally in mango[1], tomato[2] and orange.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile aroma constituents of orange.
Macleod AJ, et al
Phytochemistry, 27(7), 2185-2188 (1988)
A novel approach for nontargeted data analysis for metabolomics. Large-scale profiling of tomato fruit volatiles.
Tikunov Y, et al.
Plant Physiology, 139(3), 1125-1137 (2005)
Volatile components from mango (Mangifera indica L.) cultivars.
Pino JA, et al.
Journal of Agricultural and Food Chemistry, 53(6), 2213-2223 (2005)
Koji Uwai et al.
Bioorganic & medicinal chemistry, 16(3), 1084-1089 (2007-11-17)
Stereoselective reductive metabolism of various p-substituted acetophenone derivatives was studied using isolated rat liver 3alpha-hydroxysteroid dehydrogenase (3alpha-HSD). Kinetic experiments were performed and analyzed by measuring the products by HPLC using a chiral column. The results demonstrated that the presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service