Skip to Content
Merck
All Photos(1)

Key Documents

W262218

Sigma-Aldrich

Lavender oil 40/42% fleurs

natural, FG

Synonym(s):

Lavender oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2622
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Lavandula officinalis Chaix

Quality Level

grade

FG
Halal
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D −7°, neat

refractive index

n20/D 1.461 (lit.)

bp

204 °C (lit.)

density

0.879 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lavender; spicy; floral

Looking for similar products? Visit Product Comparison Guide

Related Categories

Preparation Note

Extraction method: steam distillation

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Yu Huang et al.
The American journal of Chinese medicine, 40(4), 845-859 (2012-07-20)
Lavender essential oil (LEO) is one the most favorite and widely used essential oils in aromatherapy. Many studies have demonstrated its functions in calming, assisting sleep, reducing pain and muscular spasms and its antiseptic function. To date, however, the mechanism
Vikas Kumar
Wiener medizinische Wochenschrift (1946), 163(3-4), 89-94 (2013-01-31)
Silexan is an essential oil produced from Lavandula angustifolia flowers with proven clinical efficacy for the treatment of anxiety disorders. The present study was conducted to assess its anxiolytic activity and to screen for neuropharmacological properties in rats and mice
P A R Hawken et al.
Physiology & behavior, 105(5), 1117-1123 (2011-12-24)
Growing concerns about the risk of addiction to benzodiazepines have led to increasing interest in alternative therapies to treat anxiety and depression. Lavender oil (Lavendula augustifolia) is reportedly anxiolytic in a number of species but little is known about how
Winai Sayorwan et al.
Journal of the Medical Association of Thailand = Chotmaihet thangphaet, 95(4), 598-606 (2012-05-23)
Investigate the effects of lavender oil on the central nervous system, autonomic nervous system, and mood responses in humans after inhalation. Twenty healthy volunteers participated in the experiments. The present study assessed autonomic parameters such as blood pressure, heart rate
Leo B Silenieks et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 20(2), 172-177 (2012-12-05)
Recently, an essential oil of selected quality produced from the flowering tops of Lavandula angustifolia Mill. by steam distillation (Silexan) has been approved in Germany for the treatment of restlessness in case of anxious mood. Based on the observed clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service