Skip to Content
Merck
All Photos(1)

Key Documents

W299200

Sigma-Aldrich

Rosemary oil

FG

Synonym(s):

Rosmarinus officinalis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2992
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
Rosemarinus officinalis L.
food allergen:
no known allergens

biological source

Rosemarinus officinalis L.

Quality Level

300
400

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 182.20

optical activity

[α]20/D +1.2°, neat

refractive index

n20/D 1.467

bp

176 °C

density

0.908 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; balsamic

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel T Organisciak et al.
Molecular vision, 19, 1433-1445 (2013-07-05)
Zinc oxide effectively reduces visual cell loss in rats exposed to intense visible light and is known to slow the rate of disease progression in advanced stages of age-related macular degeneration. Our goal was to determine the efficacy of zinc
Kanithaporn Puangsombat et al.
Journal of food science, 76(8), T174-T180 (2011-09-15)
Heterocyclic amines (HCAs) are mutagenic compounds formed when foods are cooked at high temperatures. Numerous reports have shown that natural antioxidants from spices, fruits, chocolate, and tea can inhibit formation. In this study, we evaluated HCA formation in the presence
Yang Jiang et al.
Environmental toxicology and pharmacology, 32(1), 63-68 (2011-07-27)
The composition of the essential oil of Rosemary was analyzed by gas chromatography-mass spectrometry (GC-MS). 22 components, which constitute 97.41% of the oil, were identified. The major constituents were 1,8-Cineole (26.54%) and α-Pinene (20.14%). Minimum inhibitory concentrations (MICs), minimal bactericidal
Gessilda Alcantara Nogueira de Melo et al.
Journal of medicinal food, 14(9), 944-946 (2011-06-15)
Rosmarinus officinalis L. (Lamiaceae), popularly known as rosemary, is used for food flavoring and in folk medicine as an antispasmodic, analgesic, antirheumatic, diuretic, and antiepileptic agent. Few studies have shown the anti-inflammatory effects of rosemary essential oil (REO). This study
A R Hoyle Parks et al.
Journal of animal science, 90(6), 2054-2060 (2012-06-01)
Lactic acid bacteria (LAB) can reduce Escherichia coli O157:H7 and Salmonella spp. in ground beef during storage. Furthermore, the addition of rosemary oleoresin (RO), a natural antioxidant, to ground beef has been shown to increase shelf life and is commonly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service