Skip to Content
Merck
All Photos(1)

Key Documents

W246506

Sigma-Aldrich

Eucalyptol

natural, ≥99%, FCC, FG

Synonym(s):

1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
2465
Beilstein:
105109
EC Number:
Council of Europe no.:
182
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
3.001
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥99%

refractive index

n20/D 1.457 (lit.)

bp

176-177 °C (lit.)

mp

1-2 °C (lit.)

density

0.921 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL)

Organoleptic

camphoraceous; herbaceous; minty

SMILES string

C[C@]12CC[C@H](CC1)C(C)(C)O2

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 30 ppm

Other Notes

Natural occurrence: Cardamom, cranberry, laurel, pepper, rosemary and sweet marjoram.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anke Fähnrich et al.
Plant molecular biology, 85(1-2), 135-145 (2014-02-05)
Nicotiana species of the section Alatae emit a characteristic floral scent comprising the' cineole cassette' monoterpenes 1,8-cineole, limonene, myrcene, β-pinene, α-pinene, sabinene and α-terpineol. All previously isolated 'cineole cassette'-monoterpene synthase genes are multi product enzymes that synthesize the seven compounds
Xun Hu et al.
Bioresource technology, 123, 249-255 (2012-09-04)
Bio-oil from pyrolysis of mallee (Eucalyptus loxophleba ssp. gratiae) leaves differs from that obtained with wood by its content of cyclic ethers, terpenoids and N-containing organic compounds. Upgrading of the leaf bio-oil in methanol with a solid acid catalyst was
Jossana Pereira de Sousa et al.
International journal of food microbiology, 158(1), 9-13 (2012-07-17)
This study aimed to investigate the effects of sublethal concentrations of carvacrol (CAR) and 1,8-cineole (CIN) alone and in combination on the morphology, cell viability and membrane permeability of Pseudomonas fluorescens ATCC 11253 cultivated in a vegetable-based broth. Transmission and
Hee-jin Jun et al.
Bioorganic & medicinal chemistry letters, 23(2), 579-583 (2012-12-19)
We investigated the effect of cineole on the expression of genes related to reverse cholesterol transport and hepatic fatty acid metabolism. Cineole, a small aroma compound in teas and herbs, significantly stimulated the transactivation of liver X receptor modulator (LXR)-α
Lisa A Shipley et al.
Journal of chemical ecology, 38(9), 1178-1189 (2012-10-12)
Pygmy rabbits (Brachylagus idahoensis) are one of only three vertebrates that subsist virtually exclusively on sagebrush (Artemisia spp.), which contains high levels of monoterpenes that can be toxic. We examined the mechanisms used by specialist pygmy rabbits to eliminate 1,8-cineole

Questions

  1. Can you tell us what is the amount of product present in the "Sample" size of this product? Thanks

    1 answer
    1. The SAMPLE-K quantities vary from product to product and lot-to-lot. There is no established quantity assigned. These are typically gram quantities intended for use in formulation evaluation, however some more expensive compounds may contain smaller amounts.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service