Skip to Content
Merck
All Photos(1)

Key Documents

W245000

Sigma-Aldrich

Ethyl oleate

natural, ≥85%

Synonym(s):

Oleic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
FEMA Number:
2450
Beilstein:
1727318
EC Number:
Council of Europe no.:
633
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.192
NACRES:
NA.21

biological source

palm oil

grade

Kosher
natural

Agency

meets purity specifications of JECFA

reg. compliance

FDA 21 CFR 117

Assay

≥85%

refractive index

n20/D 1.451 (lit.)

bp

216-218 °C/15 mmHg

mp

−32 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

waxy

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-

InChI key

LVGKNOAMLMIIKO-QXMHVHEDSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khushwinder Kaur
Journal of the science of food and agriculture, 95(13), 2718-2728 (2014-11-25)
Inclusion complex formation between β-cyclodextrin (β-CD) and suitable guest molecules such as cholesterol (Ch) has regularly been exploited to design self-assembled structures. In the present study an effective nanoemulsion medium (lecithin/Tween 80/ethyl oleate/water) was selected for solubilizing and stabilizing Ch
Michael D Hardy et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(27), 8187-8192 (2015-06-24)
Cell membranes are dynamic structures found in all living organisms. There have been numerous constructs that model phospholipid membranes. However, unlike natural membranes, these biomimetic systems cannot sustain growth owing to an inability to replenish phospholipid-synthesizing catalysts. Here we report
Andreas Braun et al.
Microbial cell factories, 11, 106-106 (2012-08-11)
Yarrowia lipolytica efficiently metabolizes and assimilates hydrophobic compounds such as n-alkanes and fatty acids. Efficient substrate uptake is enabled by naturally secreted emulsifiers and a modified cell surface hydrophobicity and protrusions formed by this yeast. We were examining the potential
Thomas S Muenz et al.
Die Naturwissenschaften, 99(5), 421-425 (2012-03-20)
Social work force distribution in honeybee colonies critically depends on subtle adjustments of an age-related polyethism. Pheromones play a crucial role in adjusting physiological and behavioral maturation of nurse bees to foragers. In addition to primer effects of brood pheromone
H H Orak et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 18(4), 391-402 (2012-04-24)
Antioxidant activity, colour and some nutritional properties of hot air and freeze-dried strawberry tree (Arbutus unedo L.) fruits were investigated. Additionally, the effects of two pre-treatments, namely ethyl oleate and water blanching, were compared in terms of drying characteristics. For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service