Skip to Content
Merck
All Photos(1)

Documents

W244708

Sigma-Aldrich

Ethyl nonanoate

≥98%, FCC, FG

Synonym(s):

Ethyl pelargonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOC2H5
CAS Number:
Molecular Weight:
186.29
FEMA Number:
2447
Beilstein:
1759169
EC Number:
Council of Europe no.:
388
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.107
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

0.08 mmHg ( 25 °C)

Assay

≥98%

refractive index

n20/D 1.422 (lit.)

bp

119 °C/23 mmHg (lit.)

density

0.866 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

oily; fruity; nutty

SMILES string

CCCCCCCCC(=O)OCC

InChI

1S/C11H22O2/c1-3-5-6-7-8-9-10-11(12)13-4-2/h3-10H2,1-2H3

InChI key

BYEVBITUADOIGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service