Skip to Content
Merck
All Photos(1)

Documents

W245607

Sigma-Aldrich

Ethyl propionate

≥97%, FCC, FG

Synonym(s):

2-methyl butyrate, Ethyl propanoate, Propanoic acid ethyl ester, Propionic ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOC2H5
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2456
Beilstein:
506287
EC Number:
Council of Europe no.:
402
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.121
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.52 (vs air)

vapor pressure

40 mmHg ( 27.2 °C)

Assay

≥97%

autoignition temp.

887 °F

expl. lim.

11 %

refractive index

n20/D 1.384 (lit.)

bp

99 °C (lit.)

mp

−73 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grape; ethereal; fruity; rum; sweet; wine-like

SMILES string

CCOC(=O)CC

InChI

1S/C5H10O2/c1-3-5(6)7-4-2/h3-4H2,1-2H3

InChI key

FKRCODPIKNYEAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Tuning the Electrolyte and Interphasial Chemistry for All-Climate Sodium-ion Batteries.: In a pioneering study on sodium-ion batteries, this research discusses the optimization of the electrolyte matrix, including the use of ethyl propionate, to enhance battery performance under varying climatic conditions (He et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A F Hofmann et al.
Digestive diseases and sciences, 42(6), 1274-1282 (1997-06-01)
Topical dissolution of cholesterol gallbladder stones using methyl tert-butyl ether (MTBE) is useful in symptomatic patients judged too ill for surgery. Previous studies showed that ethyl propionate (EP), a C5 ester, dissolves cholesterol gallstones rapidly in vitro, but differs from
Vibeke F Andersen et al.
The journal of physical chemistry. A, 116(21), 5164-5179 (2012-04-25)
Ethyl propionate is a model for fatty acid ethyl esters used as first-generation biodiesel. The atmospheric chemistry of ethyl propionate was investigated at 980 mbar total pressure. Relative rate measurements in 980 mbar N(2) at 293 ± 0.5 K were
O Esch et al.
Hepatology (Baltimore, Md.), 18(2), 373-379 (1993-08-01)
We performed experiments in anesthetized piglets with two cholesterol gallstone solvents, methyl tert-butyl ether and ethyl propionate, to determine whether blood levels of either solvent would increase during gallbladder instillation of these solvents under conditions simulating gallstone dissolution. The solvent
O Esch et al.
Hepatology (Baltimore, Md.), 16(4), 984-991 (1992-10-01)
Experiments were performed in anesthetized rabbits and piglets to assess gallbladder mucosal injury during irrigation with methyl tert-butyl ether, a C5 ether, or ethyl propionate, a C5 ester--two organic solvents used in the contact dissolution of cholesterol gallstones. In 44
C Y Chen et al.
Digestive diseases and sciences, 40(2), 419-426 (1995-02-01)
We evaluated the toxic effects of four currently used chemolytic solvents--dimethyl sulfoxide (DMSO, 99%), ethyl propionate (EP, 99%), tetrasodium ethyl-dimethyl tetraacetate (4Na-EDTA, 2%, pH 11), and methyl tert-butyl ether (MTBE, purity = 99.5%) in an animal model. Each solvent was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service