Skip to Content
Merck
All Photos(3)

Documents

W236411

Sigma-Aldrich

Decanoic acid

greener alternative

natural, ≥98%, FCC, FG

Synonym(s):

1-Nonanecarboxylic acid, Acid C10, C10:0, Capric acid, NSC 5025

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2364
Beilstein:
1754556
EC Number:
Council of Europe no.:
11
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.011
NACRES:
NA.21

biological source

Cocos nucifera

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.210

vapor pressure

15 mmHg ( 160 °C)

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

268-270 °C (lit.)

mp

27-32 °C (lit.)

density

0.893 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

fragrance allergen

no known allergens

greener alternative category

Organoleptic

fatty; citrus; rancid; sour

SMILES string

CCCCCCCCCC(O)=O

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)

InChI key

GHVNFZFCNZKVNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Case for supporting astrocyte energetics in glucose transporter 1 deficiency syndrome.: This review highlights the role of decanoic acid in supporting astrocyte energetics, providing a potential therapeutic approach for glucose transporter 1 deficiency syndrome and emphasizing its significance in neuroenergetics and metabolic therapies (Xu and Borges, 2024).

  • Chemical Characterization of Terpene-Based Hydrophobic Eutectic Solvents and Their Application for Pb(II) Complexation during Solvent Extraction Procedure.: This article details the development of terpene-based hydrophobic eutectic solvents incorporating decanoic acid for effective Pb(II) complexation, showcasing its potential in environmental chemistry and industrial processes (Suljkanović et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

296.6 °F - closed cup

Flash Point(C)

147 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Duran et al.
The Journal of pediatrics, 107(3), 397-404 (1985-09-01)
Five patients aged 7 to 21 months are described who developed attacks of coma after a short prodromal illness with diarrhea or vomiting or both. Four had concomitant hypoglycemia, and all had hypoketonemia, with excessive urinary excretion of medium-chain dicarboxylic
G Del Vecchio et al.
Molecular pharmaceutics, 9(9), 2523-2533 (2012-07-26)
Claudin-5 is a tight junction (TJ) protein which limits the diffusion of small hydrophilic molecules. Thus, it represents a potential pharmacological target to improve drug delivery to the tissues protected by claudin-5-dependent barriers. Sodium caprate is known as an absorption
Jürgen Zentek et al.
Archives of animal nutrition, 66(1), 14-26 (2012-03-09)
The influence of low dietary levels of free and encapsulated medium-chain fatty acids on their concentrations in the digesta, the gastric microbial ecology and bacterial metabolic products in the gastrointestinal tract (GIT) in weaned piglets was studied. Starting after weaning
Morteza Moradi et al.
Journal of chromatography. A, 1229, 30-37 (2012-02-07)
A new, efficient and environmentally friendly method for the analysis of parabens as model compounds was developed using solidified floating vesicular coacervative drop microextraction (SFVCDME). A supramolecular solvent consisting of vesicles of decanoic acid in the nano- and microscale regimes
Susanne M Krug et al.
Biomaterials, 34(1), 275-282 (2012-10-17)
Sodium caprate is a promising candidate for inducing drug absorption enhancement. The mechanism of that uptake-enhancing effect is not fully understood so far. We investigated how caprate acts in an established human intestinal cell line, HT-29/B6, on the transient opening

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service