Skip to Content
Merck
All Photos(3)

Documents

W284114

Sigma-Aldrich

2,3-Pentanedione

greener alternative

natural, ≥96%, FG

Synonym(s):

Acetylpropionyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCOCH3
CAS Number:
Molecular Weight:
100.12
FEMA Number:
2841
Beilstein:
1699638
EC Number:
Council of Europe no.:
2039c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.060
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥96%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.404 (lit.)

bp

110-112 °C (lit.)

mp

−52 °C (lit.)

density

0.957 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

caramel; creamy; buttery; sweet

storage temp.

2-8°C

SMILES string

CCC(=O)C(C)=O

InChI

1S/C5H8O2/c1-3-5(7)4(2)6/h3H2,1-2H3

InChI key

TZMFJUDUGYTVRY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Changes in physicochemical properties and microbial community succession during leaf stacking fermentation.: This research investigates the changes in physicochemical properties and microbial communities during leaf fermentation, with a focus on the role of volatile compounds like 2,3-pentanedione. The study provides insights into fermentation processes and microbial interactions (Zhang et al., 2023).

Biochem/physiol Actions

Taste at 1-5.0 ppm

Other Notes

Natural occurrence: Butter, cocoa, coffee, potato chips, roasted almonds, pecans, wheat bread and yogurt.

Legal Information

Additional information may be required prior to purchase of this material

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Sens. 1B - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - open cup

Flash Point(C)

19 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B R Epperly et al.
The Journal of biological chemistry, 264(31), 18296-18301 (1989-11-05)
Incubation of homogeneous preparations of L-threonine dehydrogenase from Escherichia coli with 2,3-butanedione, 2,3-pentanedione, phenylglyoxal, or 1,2-cyclohexanedione causes a time- and concentration-dependent loss of enzymatic activity; plots of log percent activity remaining versus time are linear to greater than 90% inactivation
G G Chang et al.
Biochimica et biophysica acta, 660(2), 341-347 (1981-08-13)
The reaction of pigeon liver malic enzyme (L-malate:NADP+ oxidoreductase (oxaloacetate-decarboxylating), EC 1.1.1.40) with dicarbonyl compounds (2,3-butanedione, methylglyoxal, 2,4-pentanedione, and phenylglyoxal) resulted in a rapid loss of its enzymatic activity. The inactivation showed pseudo-first-order kinetics for all the dicarbonyls studied. All
Synthesis and biological validation of a ubiquitous quorum-sensing molecule.
Michael M Meijler et al.
Angewandte Chemie (International ed. in English), 43(16), 2106-2108 (2004-04-15)
Roles of arginyl residues in pyridoxamine-5'-phosphate oxidase from rabbit liver.
J D Choi et al.
Biochemistry, 20(20), 5722-5728 (1981-09-29)
Magnus Rueping et al.
Organic letters, 9(5), 825-828 (2007-02-01)
[reaction: see text] A highly effective metal-catalyzed benzylation and allylic alkylation of 2,4-pentanediones has been developed. This new bismuth-catalyzed direct carbon-carbon bond forming reaction provides the corresponding monoalkylated dicarbonyl compounds in high yields after short reaction times using the lowest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service