W220500
Butyl (S)-(−)-lactate
≥97%, FG
Synonym(s):
(−)-Butyl L-lactate
About This Item
Fragrance grade
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
vapor density
4 (20 °C, vs air)
Assay
≥97%
autoignition temp.
720 °F
impurities
may contain ≤0.5% 1-Butanol (residual solvent)
bp
185-187 °C (lit.)
mp
−28 °C (lit.)
density
0.984 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
creamy; waxy; earthy
SMILES string
CCCCOC(=O)[C@H](C)O
InChI
1S/C7H14O3/c1-3-4-5-10-7(9)6(2)8/h6,8H,3-5H2,1-2H3/t6-/m0/s1
InChI key
MRABAEUHTLLEML-LURJTMIESA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
156.2 °F - closed cup
Flash Point(C)
69 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service