Skip to Content
Merck
All Photos(2)

Key Documents

389579

Sigma-Aldrich

δ-Valerolactone

technical grade

Synonym(s):

delta-Valerolactone, Tetrahydro-2H-2-pyranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
106436
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

liquid

impurities

<25% polymer

refractive index

n20/D 1.457 (lit.)

bp

226-229 °C (lit.)
58-60 °C/0.5 mmHg (lit.)

mp

−13-−12 °C (lit.)

density

1.079 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

−20°C

SMILES string

O=C1CCCCO1

InChI

1S/C5H8O2/c6-5-3-1-2-4-7-5/h1-4H2

InChI key

OZJPLYNZGCXSJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

δ-Valerolactone (tetrahydro-2H-2-pyranone or δ VL) can be used as a monomer unit in the synthesis of poly(δ-valerolactone)s poly(conjugated ester)s via ring-opening polymerization.
It can also be used as a starting material in the synthesis of (+)-guadinomic acid , sodium δ-hydroxyvalerate , methyl δ-hydroxyvalerate , and 5-hydroxyvaleraldehyde.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice E van der Ende et al.
Journal of the American Chemical Society, 130(27), 8706-8713 (2008-06-12)
We present the synthesis of discrete functionalized polyester nanoparticles in selected nanoscale size dimensions via a controlled intermolecular chain cross-linking process. The novel technique involves the controlled coupling of epoxide functionalized polyesters with 2,2'-(ethylenedioxy)bis(ethylamine) to give well-defined nanoparticles with narrow
Motofumi Osaki et al.
The Journal of organic chemistry, 74(5), 1858-1863 (2009-02-03)
BETA-cyclodextrin (beta-CD)-based nanosphere 1 initiated the oligomerization of delta-valerolactone (delta-VL) on the surface of 1 to give oligo(delta-VL)-tethered beta-CD nanosphere 2 in bulk. Atomic force microscopy indicated that the molecular size of 2 is twice that of 1. The addition
Helen Lee et al.
Biomacromolecules, 6(6), 3119-3128 (2005-11-15)
Six amphiphilic diblock copolymers based on methoxy poly(ethylene glycol) (MePEG) and poly(delta-valerolactone) (PVL) with varying hydrophilic and hydrophobic block lengths were synthesized via a metal-free cationic polymerization method. MePEG-b-PVL copolymers were synthesized using MePEG with Mn = 2000 or Mn
Faquan Zeng et al.
Bioconjugate chemistry, 17(2), 399-409 (2006-03-16)
Epidermal growth factor (EGF)-conjugated copolymer micelles were prepared from a mixture of diblock copolymers of methoxy poly(ethylene glycol)-block-poly(delta-valerolactone) (MePEG-b-PVL) and EGF-PEG-b-PVL for targeted delivery to EGF receptor (EGFR)-overexpressing cancers. The block copolymers and functionalized block copolymers were synthesized using PEG
Three-component reductive alkylation of 2-hydroxy-1, 4-naphthoquinones with lactols
Kim EE, et al.
Tetrahedron Letters, 57(8), 864-867 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service