T81108
1,3,5-Trioxane
≥99%
Synonym(s):
Metaformaldehyde, Trioxymethylene, sym.-Trioxane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
102769
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
autoignition temp.
777 °F
expl. lim.
29 %
mp
59-62 °C (lit.)
SMILES string
C1OCOCO1
InChI
1S/C3H6O3/c1-4-2-6-3-5-1/h1-3H2
InChI key
BGJSXRVXTHVRSN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
1,3,5-Trioxane is widely used as a source of anhydrous formaldehyde.
It can be used to synthesize:
It can be used to synthesize:
- Polyoxymethylene and hyperbranched polyesters.
- Calixarenes such as calix[4]resorcinarene, calix[6]resorcinarene and para-tert-butylcalix[8] and [9]arene.
- Various natural products including (−)-motuporin, (+)-sundiversifolide, (+)-lyconadin A and (−)-lyconadin B.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1 - Repr. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 1
Flash Point(F)
113.0 °F - closed cup
Flash Point(C)
45 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Single step synthesis of peripherally ?clickable? hyperbranched polyethers.
Saha A and Ramakrishnan S.
Macromolecules, 42(14), 4956-4959 (2009)
The Lyconadins: enantioselective total syntheses of (+)-lyconadin A and (−)-lyconadin B.
Beshore DC and Smith AB.
Journal of the American Chemical Society, 130(41), 13778-13789 (2008)
Hexahydro?1, 3, 5?tripropionyl?s?triazine.
Teeters WO and Gradsten MA.
Organic Syntheses, 51-51 (2003)
Enantioselective synthesis of the protein phosphatase inhibitor (−)-motuporin.
Hu T and Panek JS.
Journal of the American Chemical Society, 124(38), 11368-11378 (2002)
Cationic Copolymerization of 1, 3, 5-Trioxane with 1, 3-Dioxepane: A Comprehensive Approach to the Polyacetal Process.
Sharavanan K, et al.
Macromolecules, 42(22), 8702-8710 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service