Skip to Content
Merck
All Photos(3)

Key Documents

T80500

Sigma-Aldrich

Trimethylsulfoxonium iodide

98%

Synonym(s):

Iodotrimethyloxosulfur, S,S,S-Trimethylsulfoxonium iodide, Trimethyl(oxo)-λ6-sulfanylium iodide, Trimethyloxosulfonium iodide, Trimethyloxosulphonium iodide, Trimethylsulfoxonium iodide (7CI)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3S(I)O
CAS Number:
Molecular Weight:
220.07
Beilstein:
3595854
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

208-212 °C (dec.) (lit.)

SMILES string

[I-].C[S+](C)(C)=O

InChI

1S/C3H9OS.HI/c1-5(2,3)4;/h1-3H3;1H/q+1;/p-1

InChI key

BPLKQGGAXWRFOE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Treatment with strong base yields the ylide which adds to the carbonyl group of ketones and aldehydes to give epoxides. Also adds preferentially to the double bond of α,β-unsaturated esters to give cyclopropyl esters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 49, 5067-5067 (1993)
Tetrahedron, 48, 5089-5089 (1992)
W T Ashton et al.
Journal of medicinal chemistry, 35(11), 2103-2112 (1992-05-29)
A series of transition-state analogues having heterocyclythio C-termini has been synthesized and evaluated for inhibition of human renin. Addition of mercaptoheterocycles to a chiral Boc-amino epoxide intermediate led, after several steps, to the target [(2R,3S)-3-(BocPheHis-amino)-4-cyclohexyl-2-hydroxy-1-butyl]thio derivatives. Oxidation of the thioether
Optical phonons, crystal-field transitions, and europium luminescence-excitation processes in Eu2BaCoO5: Experiment and theory.
Taboada et al.
Physical review. B, Condensed matter, 50(13), 9157-9168 (1994-10-01)
The Journal of Organic Chemistry, 58, 3148-3148 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service