All Photos(1)
About This Item
Linear Formula:
(H2C=CHCH2)3N
CAS Number:
Molecular Weight:
137.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
4.73 (vs air)
Quality Level
vapor pressure
90 mmHg ( 80 °C)
Assay
99%
refractive index
n20/D 1.451 (lit.)
bp
150-151 °C (lit.)
density
0.79 g/mL at 25 °C (lit.)
SMILES string
C=CCN(CC=C)CC=C
InChI
1S/C9H15N/c1-4-7-10(8-5-2)9-6-3/h4-6H,1-3,7-9H2
InChI key
VPYJNCGUESNPMV-UHFFFAOYSA-N
Related Categories
Application
- Triallylamine (TAA) reacts with primary aromatic amines in the presence of a ruthenium catalyst to form 2-ethyl-3-methylquinolines.
- TAA undergoes hydrozirconation followed by transmetalation with germanium tetrachloride to form 1-aza-5-germa-5-chlorobicyclo[3.3.3]undecane. This compound can react with Grignard or lithium reagents to form the corresponding 5-organo compounds.
- The cycloaddition of TAA to fluorinated 1,3,4-oxadiazoles affords octahydro-2,7-methanofuro[3,2-c]pyridines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
87.8 °F - closed cup
Flash Point(C)
31 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ruthenium-catalyzed synthesis of 2-ethyl-3-methylquinolines from anilines and triallylamine.?
Cho C H, et al.
Tetrahedron Letters, 40(8), 1499-1500 (1999)
Palladium-catalyzed reaction of 1-aza-5-germa-5-organobicyclo [3.3. 3] undecane with aryl bromide.
Kosugi M, et al.
Journal of Organometallic Chemistry, 508(1-2), 255-257 (1996)
H G Swarts et al.
Biochimica et biophysica acta, 1189(2), 143-151 (1994-01-19)
Tertiary amines like imidazole and triallylamine lower the apparent affinity of K+ in the ATP hydrolysis reaction of pig gastric H,K-ATPase in a pH and amine concentration dependent way. The mechanism and sidedness of this effect was studied by analyzing
E D Thompson et al.
Environmental and molecular mutagenesis, 18(3), 184-199 (1991-01-01)
Three cross-linked polyacrylate polymers containing either methylenebis-acrylamide (MBA), trimethylolpropane triacrylate (TMPTA), or triallylamine (TAA) cross-linkers were tested for genotoxicity with the Salmonella mammalian microsome assay, the L5178Y mouse lymphoma TK +/- assay, the unscheduled DNA synthesis assay in primary cultures
Marc Reinholdt et al.
Membranes, 2(3), 529-552 (2012-01-01)
In the highly competitive market of fuel cells, solid alkaline fuel cells using liquid fuel (such as cheap, non-toxic and non-valorized glycerol) and not requiring noble metal as catalyst seem quite promising. One of the main hurdles for emergence of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service