Skip to Content
Merck
All Photos(2)

Key Documents

T28207

Sigma-Aldrich

Thiobenzoic acid

technical grade, 90%

Synonym(s):

Benzothioic acid, Benzoyl thiol, Monothiobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COSH
CAS Number:
Molecular Weight:
138.19
Beilstein:
2039640
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available. Contact Technical Service

grade

technical grade

Quality Level

Assay

90%

refractive index

n20/D 1.605 (lit.)

bp

122 °C/30 mmHg (lit.)

mp

15-18 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

SC(=O)c1ccccc1

InChI

1S/C7H6OS/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Yang et al.
Cancer nanotechnology, 4(4-5), 81-89 (2013-01-01)
Photothermal therapy (PTT) for cancer treatment is the use of heat between 41 and 45 °C to damage cancer cells. As a new type of transducer agent for PTT of cancer, CuS nanoparticles have several advantages. The most favorable features are
V Weber et al.
Physical chemistry chemical physics : PCCP, 17(33), 21190-21197 (2015-01-07)
Gold nanoshells, with a silica core and different core and shell dimensions, have been extensively investigated. Optical far-field properties, namely extinction and absorption, have been separately determined by means of spectrophotometry and photoacoustic spectroscopy, respectively, in the 440-900 nm range.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service