Skip to Content
Merck
All Photos(1)

Key Documents

T1199

Sigma-Aldrich

o-Toluidine solution

0.6 M in glacial acetic acid, contains thiourea as stabilizer

Synonym(s):

2-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

contains

thiourea as stabilizer

Quality Level

concentration

0.6 M in glacial acetic acid

SMILES string

Cc1ccccc1N

InChI

1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3

InChI key

RNVCVTLRINQCPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in the colorimetric determination of glucose.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Rapid specific method for determination of aldosaccharides in body fluids.
E HULTMAN
Nature, 183(4654), 108-109 (1959-01-10)
Arnaldo D'Amico et al.
Lung cancer (Amsterdam, Netherlands), 68(2), 170-176 (2009-12-05)
The use of gas sensor arrays as medical diagnosis instruments has been proposed several years ago. Since then, the idea has been proven for a limited number of diseases. The case of lung cancer is particularly interesting because it is
Tania Carreón et al.
Occupational and environmental medicine, 67(5), 348-350 (2009-11-04)
In 1991, the US National Institute for Occupational Safety and Health (NIOSH) reported an increased bladder cancer risk in a cohort of 1749 workers potentially exposed to o-toluidine and aniline at a chemical manufacturing plant. As additional information showed that
An o-toluidine method for body-fluid glucose determination.
Kurt M Dubowski
Clinical chemistry, 54(11), 1919-1920 (2008-10-30)
Kakarla Raghava Reddy et al.
Journal of nanoscience and nanotechnology, 7(9), 3117-3125 (2007-11-21)
Poly(ortho-toluidine) (POT)-gold (Au) and palladium (Pd) composite nanospheres were successfully synthesized by the reaction of o-toluidine with the corresponding metal (Au or Pd) colloidal solution through self-assembly process in the presence of dodecylbenzenesulfonic acid (DBSA), which acts as both a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service