Skip to Content
Merck
All Photos(3)

Key Documents

T109

Sigma-Aldrich

L-(+)-Tartaric acid

≥99.5%

Synonym(s):

(2R,3R)-(+)-Tartaric acid, L-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725147
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

5.18 (vs air)

Quality Level

Assay

≥99.5%

form

granular
powder or crystals

optical activity

[α]20/D +13.5±0.5°, c = 10% in H2O

autoignition temp.

797 °F

mp

170-172 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m1/s1

InChI key

FEWJPZIEWOKRBE-JCYAYHJZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-(+)-tartaric acid is an organic compound that is commonly used as a chiral auxiliary in reactions that involve the addition of a chiral nucleophile. It is also used as a resolving agent for the separation of enantiomers from a racemic mixture. Additionally, L-(+)-tartaric acid can also be used as a building block for the synthesis of other chiral compounds.

Application

L- (+)-Tartaric acid can be used as:

  • A co-former for the synthesis of etravirine co-crystals.
  • A mobile phase additive in thin-layer chromatography.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Growth, molecular structure, NBO analysis and vibrational spectral analysis of l-tartaric acid single crystal.
Sasikala, V .et al.
Spectrochimica Acta Part A: Molecular Spectroscopy, 123, 127-141 (2014)
Amalia Papadaki et al.
Bio-protocol, 9(19), e3384-e3384 (2019-10-05)
Acid ecto-phosphatases are enzymes that hydrolyze phosphomonoesters in the acidic pH range with their active sites facing the extacellular medium. Their activities can be measured in living cells. In bacteria and protozoan pathogens, acid ecto-phosphatases have been associated with the
Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different
Mark D Eddleston et al.
Chemical communications (Cambridge, England), 48(92), 11340-11342 (2012-10-18)
The formation of diastereomeric cocrystals of malic acid and tartaric acid was investigated by liquid-assisted grinding in the solid state. We demonstrate that racemic malic acid can be converted into two distinct diastereomeric cocrystal phases by grinding with a single
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service