Skip to Content
Merck
All Photos(4)

Key Documents

S8876

Sigma-Aldrich

Sulfamerazine

ReagentPlus®, ≥99.0%

Synonym(s):

4-Amino-N-(4-methyl-2-pyrimidinyl)benzenesulfonamide, N1-(4-Methylpyrimidin-2-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O2S
CAS Number:
Molecular Weight:
264.30
Beilstein:
249133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

≥99.0%

SMILES string

Cc1ccnc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C11H12N4O2S/c1-8-6-7-13-11(14-8)15-18(16,17)10-4-2-9(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)

InChI key

QPPBRPIAZZHUNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Pan et al.
Chemical communications (Cambridge, England), 47(1), 352-354 (2010-08-24)
A phosphine-mediated one-step disulfide formation from S-nitrosothiols has been developed. This reaction can convert unstable S-nitrosothiols to stable disulfides via sulfenamide intermediates under very mild conditions. It has the potential to be used for the detection of S-nitrosothiols.
M S P De Lima et al.
Carbohydrate research, 344(13), 1709-1715 (2009-06-27)
In this work chitosan membranes modified by contact with poly(acrylic acid) (PAA) aqueous solution at two different temperatures (25 degrees C and 60 degrees C) were obtained. The pure chitosan (CS) membranes, as well as those treated with PAA (CSPAA_25
Xian Zhang et al.
Environmental science and pollution research international, 19(5), 1392-1404 (2012-06-30)
Estrogenic compounds and antibiotic residues in environment are receiving significant attention because of their potential adverse effects on ecosystems and human health. The objectives of this study were to determine the occurrence and seasonal variability of eight kinds of estrogenic
Regioselective hydroxysulfenylation of alpha,beta-unsaturated imines: enhanced stability of an intermediate radical.
Masafumi Ueda et al.
Angewandte Chemie (International ed. in English), 47(30), 5600-5604 (2008-06-17)
A practical sulfenylation of 2,5-diketopiperazines.
K C Nicolaou et al.
Angewandte Chemie (International ed. in English), 51(3), 728-732 (2011-12-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service