Skip to Content
Merck
All Photos(1)

Key Documents

S4379

Sigma-Aldrich

Sodium phosphate glass

Type 45

Synonym(s):

sodium orthophosphate glass

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

Pricing and availability is not currently available.

Quality Level

mol wt

45 P±5 P (av. chain length)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PZ0184PZ0333SML2809
WNK463 ≥98% (HPLC)

SML2809

WNK463

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

solubility

DMSO: >30 mg/mL

solubility

DMSO: 20 mg/mL

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to off-white

color

white to beige

color

white to beige

color

white to beige

General description

Sodium phosphate glass is composed of Na2O–P2O5. It is used as a sequestering agent in hard water treatment and also as a dispersant in clay processing and pigment production.[1] In Na-ion batteries, sodium phosphate glass can be used as a solid electrolyte.[2]

Application

Sodium phosphate glass type 45 (polyP45) can be used as a reference material in the extraction and quantification of inorganic polyphosphate residues found in microorganisms.[3][4]

Biochem/physiol Actions

Reported to be of use in protein binding and sequestering and in studies on bone calcification.

Other Notes

Nan+2PnO3n+1; n = the average number of phosphorus (P) atoms in the chain.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    An improved method for extraction and quantification of polyphosphate granules from microbial cells
    Mukherjee C and Ray K
    Protocol Exchange, 10 (2015)
    Extraction and quantification of polyphosphate in the budding yeast Saccharomyces cerevisiae
    Canadell D, et al.
    Bio-protocol, 6(14), e1874-e1874 (2016)
    Huilong Ou et al.
    Microorganisms, 8(1) (2020-01-08)
    Some sponges have been shown to accumulate abundant phosphorus in the form of polyphosphate (polyP) granules even in waters where phosphorus is present at low concentrations. But the polyP accumulation occurring in sponges and their symbiotic bacteria have been little
    An improved method for extraction and quantification of polyphosphate granules from microbial cells
    Mukherjee C and Ray K
    Protocol Exchange, 10 (2015)
    The structure of simple phosphate glasses
    Brow RK
    Journal of Non-Crystalline Solids, 263, 1-28 (2000)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service