305553
Sodium hexametaphosphate
crystalline, +200 mesh, 96%
Synonym(s):
SHMP, Calgon, Phosphate glass, water soluble, Polyphosphate sodium salt, Sodium polyphosphate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
form:
crystalline
Recommended Products
Quality Level
Assay
96%
form
crystalline
particle size
+200 mesh
SMILES string
[Na+].[Na+].[P](=O)([O-])([O-])O[P](=O)(O)O
InChI
1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2
InChI key
GYQBBRRVRKFJRG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Sodium hexametaphosphate is an inorganic polyphosphate salt commonly used as a corrosion inhibitor, emulsifying agent and as a tooth whitening agent in dentifrice formulations.
Sodium hexametaphosphate is a water-soluble inorganic stabilizing agent used as a surfactant to decrease the zeta potential of halloysite suspensions.
Sodium hexametaphosphate is a water-soluble inorganic stabilizing agent used as a surfactant to decrease the zeta potential of halloysite suspensions.
Application
Sodium hexametaphosphate has been used as a polyphosphate anion source to prepare hydrated calcium polyphosphate powder. It can be used as a deflocculant to prepare suspensions of kaolinite and ceramic grade kaolin.
Biochem/physiol Actions
Intended for the cytologic demonstration of Naphthol AS-D Chloroacetate Esterase in blood, bone marrow films or tissue touch preparations.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dispersion of phyllosilicates in aqueous suspensions: Role of the nature and amount of surfactant
Nadia H et al.
Journal of Colloid and Interface Science, 425, 67-74 (2014)
Amy M Kahler et al.
Water, 7(3), 1202-1216 (2015-11-05)
In this study, hollow-fiber ultrafiltration (UF) was assessed for recovery of
Synthesis of BaSO4 nanoparticles by precipitation method using sodium hexa metaphosphate as a stabilizer
Asha G et al.
Solid State Communications, 150, 386-388 (2010)
Amorphous calcium phosphate powder synthesized from calcium acetate and polyphosphoric acid for bioceramics application.
Safronova T V, et al.
Ceramics International, 43(1), 1310-1317 (2017)
The role of sodium hexametaphosphate in the dissolution process of kaolinite and kaolin.
Andreola F, et al.
J. Eur. Ceram. Soc., 24(7), 2113-2124 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service