R2252
Rubidium chloride
ReagentPlus®, ≥99.0% (metals basis)
Synonym(s):
Rubidium monochloride, Rubidium(I) chloride
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
reagent
form:
powder
solubility:
H2O: soluble, clear to slightly hazy
Recommended Products
grade
reagent
product line
ReagentPlus®
Assay
≥99.0% (metals basis)
form
powder
impurities
≤10000 ppm Metal Trace Analysis (ICP)
mp
715 °C (lit.)
solubility
H2O: soluble, clear to slightly hazy
density
2.8 g/mL at 25 °C (lit.)
absorption
≤0.4 at 280 in H2O at 50%
SMILES string
[Cl-].[Rb+]
InChI
1S/ClH.Rb/h1H;/q;+1/p-1
InChI key
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Rubidium chloride is an alkali metal halide saltand an excellent water-soluble crystalline rubidium source. It is widely used in thefield of biomedical research, electrochemistry, nanomaterial synthesis, and thepetrol industry.
Application
Rubidium chloride can be used as a dopant to synthesize magnesium oxide nanoparticles.
It can be used to enhance the performance of OLEDs. For example, a thin layer of RbCl inserted into a multilayer OLED (ITO/NPB/Alq3/Al) can significantly increase its electroluminescent efficiency. This can happen due to the lowering of the electron injection barrier and enhancing the recombination rate induced by trap sites provided by the RbCl layer.
Rubidium chloride (RbCl) can also be used as a non-invasive biomarker.
It may also be used to study the chronotypic effects of Rb in hamsters.
It can be used to enhance the performance of OLEDs. For example, a thin layer of RbCl inserted into a multilayer OLED (ITO/NPB/Alq3/Al) can significantly increase its electroluminescent efficiency. This can happen due to the lowering of the electron injection barrier and enhancing the recombination rate induced by trap sites provided by the RbCl layer.
Rubidium chloride (RbCl) can also be used as a non-invasive biomarker.
It may also be used to study the chronotypic effects of Rb in hamsters.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rubidium chloride fuses split circadian activity rhythms in hamsters housed in bright constant light
Hallonquist J, et al.
Chronobiology International, 11(2), 65-71 null
Marking bark beetle parasitoids within the host plant with rubidium for dispersal studies
Hougardy E, et al.
Entomologia Experimentalis et Applicata, 108(2), 107-114 (2003)
The effect of rubidium chloride on properties of organic light-emitting diodes
Lu Z, et al.
Solid-State Electron, 53(11), 1154-1158 (2009)
Balasundaresan Dhakshnamoorthy et al.
Journal of molecular biology, 396(2), 293-300 (2009-11-26)
The OmpF porin from the Escherichia coli outer membrane folds into a trimer of beta-barrels, each forming a wide aqueous pore allowing the passage of ions and small solutes. A long loop (L3) carrying multiple acidic residues folds into the
Amer Alam et al.
Nature structural & molecular biology, 16(1), 30-34 (2008-12-23)
We report the crystal structure of the nonselective cation channel NaK from Bacillus cereus at a resolution of 1.6 A. The structure reveals the intracellular gate in an open state, as opposed to the closed form reported previously, making NaK
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service