Skip to Content
Merck
All Photos(1)

Documents

251445

Sigma-Aldrich

Rubidium iodide

99.9% trace metals basis

Synonym(s):

Hydriodic acid rubidium salt, Rubidium monoiodide, Rubidium(1+) iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbI
CAS Number:
Molecular Weight:
212.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

powder

impurities

≤1500.0 ppm Trace Metal Analysis

bp

1300 °C/1 atm (lit.)

mp

656 °C (lit.)

solubility

ethanol: soluble(lit.)

density

3.55 g/mL at 25 °C (lit.)

SMILES string

[Rb+].[I-]

InChI

1S/HI.Rb/h1H;/q;+1/p-1

InChI key

WFUBYPSJBBQSOU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man Singh et al.
Chemistry & biodiversity, 2(6), 809-824 (2006-12-29)
Density (rho), apparent molar volume (V(phi)), and viscosity (eta) of 0.0010 to 0.0018% (w/v) of bovine serum albumin (BSA), egg albumin, and lysozyme in 0.0002, 0.0004, and 0.0008 M aqueous RbI and CsI, and (dodecyl)(trimethyl)ammonium bromide (DTAB) solutions were obtained.
Contact dermatitis from rubidium iodide in eyedrops.
N Cameli et al.
Contact dermatitis, 23(5), 377-378 (1990-11-01)
Johan Mähler et al.
Inorganic chemistry, 51(1), 425-438 (2011-12-16)
The hydration of the alkali metal ions in aqueous solution has been studied by large angle X-ray scattering (LAXS) and double difference infrared spectroscopy (DDIR). The structures of the dimethyl sulfoxide solvated alkali metal ions in solution have been determined
P K Manimozhi et al.
Luminescence : the journal of biological and chemical luminescence, 22(5), 468-472 (2007-07-05)
Optical absorption, photoluminescence, thermoluminescence (TL) and photostimulated luminescence (PSL) studies on RbI:Tb(3+) crystals irradiated with gamma-rays is reported. Photoluminescence of these crystals exhibits characteristic Tb(3+) emissions, due to transitions from the (5)D(3) and (5)D(4) levels to various levels of the
Heping Shen et al.
Science advances, 4(12), eaau9711-eaau9711 (2018-12-18)
Increasing the power conversion efficiency of silicon (Si) photovoltaics is a key enabler for continued reductions in the cost of solar electricity. Here, we describe a two-terminal perovskite/Si tandem design that increases the Si cell's output in the simplest possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service