Skip to Content
Merck
All Photos(1)

Key Documents

QBD10918

Sigma-Aldrich

m-dPEG®48-amine

>95% (HPLC)

Synonym(s):

m-PEG48-amine, mPEG-amine, mPEG2000-amine, mPEG48-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C97H197NO48
Molecular Weight:
2145.58
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC

InChI key

NCWDZRVHWBSGGG-UHFFFAOYSA-N

Features and Benefits

m-dPEG48-amine is a methyl-terminated PEG amine compound. The PEG is a water-soluble, single molecular weight PEG product with a discrete chain length (dPEG). This dPEG linker is 146 atoms (173.1 Å) long, a monodisperse equivalent of a PEG2000. Carboxylic acids and their active esters react with the amino terminus to form amide bonds. Also, the amino terminus reacts with aldehydes and ketones, forming Schiff bonds, which are reducible to secondary amines. Uses for m-dPEG48-amine include modification of acid-functionalized surfaces and free carboxylic acid groups on biomolecules.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George Badescu et al.
Bioconjugate chemistry, 25(3), 460-469 (2014-02-12)
Many clinically used protein therapeutics are modified to increase their efficacy. Example modifications include the conjugation of cytotoxic drugs to monoclonal antibodies or poly(ethylene glycol) (PEG) to proteins and peptides. Monothiol-specific conjugation can be efficient and is often accomplished using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service