Skip to Content
Merck
All Photos(1)

Key Documents

JKA7032

Sigma-Aldrich

4arm-PEG2K

average MN 2,000, NH2, carboxyl reactive

Synonym(s):

4arm-PEG-NH2, 4arm-PEG-amine, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C(CH2O(CH2CH2O)nCH2CH2NH2)4
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

4arm-PEG2K-NH2, average Mn 2,000

form

liquid

mol wt

average Mn 2,000

reaction suitability

reactivity: carboxyl reactive

polymer architecture

shape: 4-arm
functionality: homofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

4arm PEG Amine. Hydrogel PEG. Amine group binds to carboxylic group (-COOH) or other amine reactive chemical groups.

Legal Information

Product of JenKem Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

McKinnon, D.D., et al.
Chemistry of Materials, 26(7) (2014)
Maitz, M.F., et al.
Nature Communications, 4 (2013)
Xu, J., et al.
Journal of the American Chemical Society, 808 (2014)

Articles

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service