Skip to Content
Merck
All Photos(2)

Key Documents

P38005

Sigma-Aldrich

1,3,5-Trihydroxybenzene dihydrate

97%

Synonym(s):

Phloroglucinol dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3 · 2H2O
CAS Number:
Molecular Weight:
162.14
Beilstein:
5109263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

218-221 °C (A) (lit.)

SMILES string

O.O.Oc1cc(O)cc(O)c1

InChI

1S/C6H6O3.2H2O/c7-4-1-5(8)3-6(9)2-4;;/h1-3,7-9H;2*1H2

InChI key

MPYXTIHPALVENR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

In microscopy as a decalcifier of bone specimens. Reagent for pentoses, pentosans and aldehydes.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Margaret Murray et al.
Antioxidants (Basel, Switzerland), 8(2) (2019-03-01)
When healthy adults consume carbohydrates at night, postprandial blood glucose responses are elevated and prolonged compared to daytime.Extended postprandial hyperglycaemia is a risk factor for type 2 diabetes. Polyphenols are bioactive secondary metabolites of plants and algae with potential to
Margaret Murray et al.
Nutrients, 10(3) (2018-03-03)
This study investigated the impact of a polyphenol-rich seaweed extract on postprandial glycaemia in healthy adults, and, as a secondary outcome, the influence of ethnicity on these outcomes. Thirty-eight volunteers (26 non-Asian, 12 Asian) aged 19 to 56 years participated
Yan Wang et al.
Journal of separation science, 42(7), 1332-1340 (2019-01-23)
Three monomers, octakis (3-mercaptopropyl) octasilsesquioxane, 1,2,4-trivinylcyclohexane and isophytol were employed to synthesize a novel monolithic stationary phase via photo-initiated thiol-ene click polymerization for reversed-phase liquid chromatography. Several factors such as porogenic system, reaction time and the molar ratio of functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service