Skip to Content
Merck
All Photos(1)

Key Documents

79330

Sigma-Aldrich

Phloroglucinol

≥99.0% (HPLC)

Synonym(s):

1,3,5-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
1341907
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

solid

impurities

diresorcin, none detected
≤2% water

mp

215-220 °C

SMILES string

Oc1cc(O)cc(O)c1

InChI

1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H

InChI key

QCDYQQDYXPDABM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Phloroglucinol (phlo) is a phenol derivative that shows cyctoprotective effect from oxidative damage by enhancing the activity of cellular catalase.
  • It can react with benzaldehyde derivatives to form phloroglucinol-based microporous polymeric organic frameworks (phlo-POF) with potential applications in ion-exchange and gas adsorption.
  • Phlo can also be used to prepare synthetic analogs of A-type proanthocyanidins (PACs) such as 2,8-dioxabicyclo[3.3.1]nonane derivatives by reacting with the corresponding flavylium salts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Birkemeyer et al.
Metabolites, 10(9) (2020-09-17)
Accumulation of biologically active metabolites is a specific feature of plant biochemistry, directing the use of plants in numerous applications in the pharmaceutical and food industries. Among these substances, the plethora of phenolic compounds has attracted particular interest among researchers.
Thermodynamic Stability of Flavylium Salts as a Valuable Tool To Design the Synthesis of A-Type Proanthocyanidin Analogues
Alejo-Armijo A, et al.
The Journal of Organic Chemistry, 83(19), 12297-12304 (2018)
Efficient and Rapid Photocatalytic Reduction of Hexavalent Chromium Achieved by a Phloroglucinol-Derived Microporous Polymeric Organic Framework Solid
Kostas V, et al.
The Journal of Physical Chemistry C, 121(13), 7303-7311 (2017)
Anne M Vissers et al.
Phytochemical analysis : PCA, 28(6), 487-495 (2017-06-15)
Phlorotannins are complex mixtures of phloroglucinol oligomers connected via C-C (fucols) or C-O-C (phlorethols) linkages. Their uniformity in subunits and large molecular weight hamper their structural analysis. Despite its commercial relevance for alginate extraction, phlorotannins in Laminaria digitata have not
Cytoprotective effect of phloroglucinol on oxidative stress induced cell damage via catalase activation
Kang KA, et al.
Journal of Cellular Biochemistry, 97(3), 609-620 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service