Skip to Content
Merck
All Photos(1)

Documents

P27100

Sigma-Aldrich

N-Phenylmaleimide

97%

Synonym(s):

1-Phenyl-pyrrole-2,5-dione, 1-[4-(Acetoxy)phenyl]maleimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
125098
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

162-163 °C/12 mmHg (lit.)

mp

85-87 °C (lit.)

SMILES string

O=C1C=CC(=O)N1c2ccccc2

InChI

1S/C10H7NO2/c12-9-6-7-10(13)11(9)8-4-2-1-3-5-8/h1-7H

InChI key

HIDBROSJWZYGSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vânia F Noldin et al.
Pharmacological reports : PR, 63(3), 772-780 (2011-08-23)
Myeloperoxidase (MPO) is an important enzyme that catalyzes the reaction between hydrogen peroxide and chloride to generate hypochlorous acid, which oxidizes a range of biomolecules and has been associated with inflammatory diseases. The synthetic compounds N-phenylmaleimide (NFM) and 4-methyl-N-phenylmaleimide (Me-NFM)
Jian-Hui Wen et al.
Talanta, 84(1), 13-18 (2011-02-15)
Representing a compound by a numerous structural descriptors becomes common in quantitative structure-activity relationship (QSAR) studies. As every descriptor carries molecular structure information more or less, it seems more advisable to investigate all the possible descriptor vectors rather than traditional
Masato Hoshino et al.
Chemical & pharmaceutical bulletin, 56(4), 480-484 (2008-04-02)
Diels-Alder reactions between N-phenylmaleimide, acting as the dienophile, and 2(1H)-pyridones having a methoxy or a chloro substituent, were carried out, under atmospheric and high pressure conditions, to give the corresponding isoquinuclidine derivatives. Stereoselectivity of the Diels-Alder reactions was studied using
Nicolas Matuszak et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 45(3), 263-271 (2011-12-01)
The endocannabinoid system is implicated in numerous physiopathological processes while more and more pieces of evidence wave the link between this complex machinery and cancer related phenomenon. In these lines, we confirmed the effects of 2-arachidonoylglycerol (2-AG), the main endocannabinoid
Michael L Vasil et al.
Antimicrobial agents and chemotherapy, 56(12), 6223-6234 (2012-09-26)
The twin-arginine translocase (TAT) in some bacterial pathogens, including Pseudomonas aeruginosa, Burkholderia pseudomallei, and Mycobacterium tuberculosis, contributes to pathogenesis by translocating extracellular virulence determinants across the inner membrane into the periplasm, thereby allowing access to the Xcp (type II) secretory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service