P25485
D−(−)-α-Phenylglycine
99%, detection
Synonym(s):
(R)-(−)-2-Phenylglycine, D-2-Phenylglycine, R-(−)-α-Aminophenylacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH(NH2)CO2H
CAS Number:
Molecular Weight:
151.16
Beilstein:
2208676
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Product Name
D−(−)-α-Phenylglycine, 99%
Quality Level
Assay
99%
form
powder or crystals
optical activity
[α]20/D −155°, c = 1 in 1 M HCl
reaction suitability
reaction type: solution phase peptide synthesis
color
white
mp
302 °C (dec.) (lit.)
application(s)
detection
SMILES string
N[C@@H](C(O)=O)c1ccccc1
InChI
1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)/t7-/m1/s1
InChI key
ZGUNAGUHMKGQNY-SSDOTTSWSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
302.0 °F - closed cup
Flash Point(C)
150 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Martijn A Hoeben et al.
Biotechnology and bioengineering, 93(4), 607-617 (2006-01-06)
The interfacial partitioning behavior of ampicillin and phenylglycine crystals in different two-phase systems has been investigated. The two-phase systems employed are water/dodecane, water/1-butanol, and water/pentane/methanol. By means of partition experiments and microscopic imaging, it has been shown that the mechanism
Choedchai Saehuan et al.
Biochimica et biophysica acta, 1770(11), 1585-1592 (2007-10-06)
Following induction with D-phenylglycine both d-phenylglycine aminotransferase activity and benzoylformate decarboxylase activity were observed in cultures of Pseudomonas stutzeri ST-201. Induction with benzoylformate, on the other hand, induced only benzoylformate decarboxylase activity. Purification of the benzoylformate decarboxylase, followed by N-terminal
Caroline Haurena et al.
The Journal of organic chemistry, 75(8), 2645-2650 (2010-03-23)
A range of alpha-amino esters has been synthesized in good to high yields using a straightforward three-component reaction among preformed or in situ generated aromatic or benzylic organozinc reagents, primary or secondary amines, and ethyl glyoxylate. The procedure, which is
Kuoxi Xu et al.
Chirality, 24(8), 646-651 (2012-05-24)
The triazine-based bisbinaphthyl crown ethers oxacalix[2]arene[2]bisbinaphthes R-1, R-2, R-3 and S-1, S-2, S-3 were synthesized. The interactions of these compounds with various α-aminocarboxylic acid anions were studied. The crown ethers were found to carry out highly enantioselective fluorescent recognition of
Hyung Min Kim et al.
The Journal of chemical physics, 128(18), 184313-184313 (2008-06-06)
We investigated the conformational structures of L-phenylglycine in the gas phase by photoionization and double resonance spectroscopy techniques as well as high-level ab initio calculations. The UV-UV and IR-UV double resonance spectroscopy suggested that there exists only one conformer that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service