Skip to Content
Merck
All Photos(2)

Documents

P14815

Sigma-Aldrich

4-Ethoxyaniline

98%

Synonym(s):

p-Phenetidine, 4-Ethoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4NH2
CAS Number:
Molecular Weight:
137.18
Beilstein:
606666
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.559 (lit.)

bp

250 °C (lit.)

mp

2-5 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccc(N)cc1

InChI

1S/C8H11NO/c1-2-10-8-5-3-7(9)4-6-8/h3-6H,2,9H2,1H3

InChI key

IMPPGHMHELILKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J D Baty et al.
Journal of chromatography, 353, 329-337 (1986-02-26)
Liquid chromatographic methods were developed for the study of the in vitro acetylation of the sulphonamide drug sulphamethazine and a series of aniline derivatives. The sensitivity of the methods have allowed data on the activity of the N-acetyltransferase enzyme(s) in
Esko Kankuri et al.
Thrombosis research, 110(5-6), 299-303 (2003-11-01)
The present study was aimed to test the possible cyclooxygenase (COX)-1/COX-2 selectivity of the old analgesic drug phenacetin and its metabolite p-phenetidine, which exhibits high renal toxicity. Paracetamol (acetaminophen), the main metabolite of phenacetin with low renal toxicity, and indomethacin
T Lindqvist et al.
Chemical research in toxicology, 4(4), 489-496 (1991-07-01)
4-Ethoxyaniline (p-phenetidine) is oxidized by peroxidases to form several products, one of which is 4-[(4-ethoxyphenyl)imino]-2,5-cyclohexadien-1-one (1). This compound reacts with N-acetylcysteine (NAC) in methanol-phosphate buffers, generating at least four different products. Four major products, 4-[(4-ethoxyphenyl)amino]phenol (2), 3-(N-acetylcystein-S-yl)-4-[(4-ethoxyphenyl)amino]phenol (3), 2,5-bis(N-acetylcystein-S-yl)-4-[(4-ethoxyphenyl)-amino]phenol (4)
R Larsson et al.
The Journal of pharmacology and experimental therapeutics, 235(2), 475-480 (1985-11-01)
The metabolism of p-phenetidine in microsomes from rabbit kidney and the metabolism of acetaminophen and p-phenetidine in human kidney microsomes to protein binding metabolites were examined. Microsomal preparations from rabbit kidney medulla catalyzed the irreversible arachidonic acid-dependent binding of p-[14C]phenetidine
Annabel Kuek et al.
Postgraduate medical journal, 83(978), 251-260 (2007-04-04)
Targeted biologic therapies have revolutionised treatment of immune-mediated inflammatory diseases (IMIDs) due to their efficacy, speed of onset and tolerability. The discovery that clinically unrelated conditions, such as rheumatoid arthritis and Crohn's disease, share similar immune dysregulation has led to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service