Skip to Content
Merck
All Photos(2)

Documents

N7524

Sigma-Aldrich

Bis(cyclopentadienyl)nickel(II)

Synonym(s):

Di(cyclopentadienyl)nickel(II), Nickelocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(C5H5)2
CAS Number:
Molecular Weight:
188.88
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Ni, 20.0-31.5% EDTA titration

reaction suitability

core: nickel
reagent type: catalyst

mp

171-173 °C (lit.)

SMILES string

[Ni].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Ni/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

FOOKRXHSBKEWSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This metal alkyl may be used to synthesize hybrid organic-inorganic polymeric molecular layer deposition (MLD). Bis(cyclopentadienyl) nickel (Cp 2 Ni) may be used as a Ni precursor for the atomic layer deposition (ALD) of NiO/carbon nanotubes hybrids.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1A - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced Catalytic Activity for Methanol Electro-oxidation of Uniformly Dispersed Nickel Oxide Nanoparticles?Carbon Nanotube Hybrid Materials.
Tong X, et al.
Small, 8(22), 3390-3395 (2012)
Molecular Layer Deposition of Organic and Hybrid Organic-Inorganic Polymers.
George SM and Yoon B
Material Matters, 3(2), 34-37 null
Muhammad Hamid Raza et al.
ACS applied materials & interfaces, 12(4), 4594-4606 (2020-01-15)
The gas-sensing properties and mechanism and the role of the shell thickness of structurally well-defined SnO2/NiO heterostructures are studied. One-dimensional (1D) SnO2/NiO core-shell nanowires (CSNWs) were produced by a two-step process; single-crystalline SnO2-core nanowires (NWs) were synthesized by vapor-liquid-solid (VLS)
Patrick P Komane et al.
International journal of molecular sciences, 21(7) (2020-03-29)
Currently, there is a lack of ultrasensitive diagnostic tool to detect some diseases such as ischemic stroke, thereby impacting effective and efficient intervention for such diseases at an embryonic stage. In addition to the lack of proper detection of the

Articles

Atomic layer deposition meets various needs including semiconductor device miniaturization and nanoparticle coating.

Atomic layer deposition meets various needs including semiconductor device miniaturization and nanoparticle coating.

Atomic layer deposition meets various needs including semiconductor device miniaturization and nanoparticle coating.

Atomic layer deposition meets various needs including semiconductor device miniaturization and nanoparticle coating.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service