Skip to Content
Merck
All Photos(1)

Key Documents

M1407

Sigma-Aldrich

Malononitrile

≥99%

Synonym(s):

Dicyanomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(CN)2
CAS Number:
Molecular Weight:
66.06
Beilstein:
773697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

bp

220 °C (lit.)

mp

30-32 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

N#CCC#N

InChI

1S/C3H2N2/c4-2-1-3-5/h1H2

InChI key

CUONGYYJJVDODC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malononitrile, a weak cyanocarbon acid, is a versatile compound with exceptional reactivity. This crystalline aliphatic nitrile is used as a building block to synthesize heterocyclic compounds and polymers

Application

Malononitrile may be used in the:
  • base-promoted on-water synthesis of [1,6]-naphthyridines.†
  • synthesis of γ-ketoamides.
  • preparation of heterocyclic privileged medicinal scaffolds involving pyridine, 1,4-dihydropyridine, chromeno[2,3-b]pyridine and dihydro-1,4-dithiepine frameworks.

Packaging

Packaged in glass bottles

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Performance Quinoline-Malononitrile Core as a Building Block for the Diversity-Oriented Synthesis of AIEgens
Z Guo, et al.
Angewandte Chemie (International Edition in English), 59, 9812-9825 (2020)
The chemistry of malononitrile.
F Freeman
Chemical reviews, 69(5), 591-624 (1969-10-01)
Kamila K Mentel et al.
Nature communications, 9(1), 2903-2903 (2018-07-27)
Electron transfer reactions are arguably the simplest chemical reactions but they have not yet ceased to intrigue chemists. Charge-separation and charge-recombination reactions are at the core of life-sustaining processes, molecular electronics and solar cells. Intramolecular electron donor-acceptor systems capture the
Enxiang Wei et al.
Organic & biomolecular chemistry, 12(33), 6389-6392 (2014-07-11)
An efficient synthesis of γ-ketoamides was developed by the one-pot multicomponent reaction of chalcones, malononitrile and DMF (as both the reactant and solvent) in the presence of NaOH (3.0 equiv.). The reaction features high atom economy, easily available starting materials
Nikolai M Evdokimov et al.
The Journal of organic chemistry, 72(9), 3443-3453 (2007-04-06)
Heterocyclic privileged medicinal scaffolds involving pyridine, 1,4-dihydropyridine, chromeno[2,3-b]pyridine, and dihydro-1,4-dithiepine frameworks are prepared via a single-step multicomponent reaction of structurally diverse aldehydes with various thiols and malononitrile. Mechanistic studies of the synthetic pathway leading to pyridines reveal that 1,4-dihydropyridines undergo

Articles

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service