Skip to Content
Merck
All Photos(5)

Key Documents

M81705

Sigma-Aldrich

Dimethyl sulfone

98%

Synonym(s):

Methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO2
CAS Number:
Molecular Weight:
94.13
Beilstein:
1737717
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

bp

238 °C (lit.)

mp

107-109 °C (lit.)

SMILES string

CS(C)(=O)=O

InChI

1S/C2H6O2S/c1-5(2,3)4/h1-2H3

InChI key

HHVIBTZHLRERCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethyl sulfone (DMSO2) can undergo:
  • Acylation with N-acylbenzotriazoles to synthesize β-keto sulfones.
  • α-Monoarylation via palladium-catalyzed Negishi coupling reaction to synthesize benzylic sulfones.
  • One-step olefination of alcohols in the presence of ruthenium pincer complex as a catalyst.

DMSO2 can also be used as an electrolyte over a wide range of temperatures (110–150°C) for electrodeposition of aluminum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct catalytic olefination of alcohols with sulfones.
Srimani D, et al.
Angewandte Chemie (International ed. in English), 53(41), 11092-11095 (2014)
Efficient conversion of sulfones intoβ-keto sulfones by N-acylbenzotriazoles.
Katritzky AR, et al.
The Journal of Organic Chemistry, 68(4), 1443-1446 (2003)
Studies on the AlCl3/dimethylsulfone (DMSO2) electrolytes for the aluminum deposition processes.
Jiang T, et al.
Surface and Coatings Technology, 201(14), 6309-6317 (2007)
Eytan M Debbi et al.
BMC complementary and alternative medicine, 11, 50-50 (2011-06-29)
Patients with osteoarthritis (OA) take a variety of health supplements in an attempt to reduce pain and improve function. The aim of this study was to determine the efficacy of methylsulfonylmethane (MSM) in treating patients with knee OA. This study
Eun Joung Lim et al.
PloS one, 7(4), e33361-e33361 (2012-04-10)
Breast cancer is the most aggressive form of all cancers, with high incidence and mortality rates. The purpose of the present study was to investigate the molecular mechanism by which methylsulfonylmethane (MSM) inhibits breast cancer growth in mice xenografts. MSM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service