Skip to Content
Merck
All Photos(1)

Key Documents

K2755

Sigma-Aldrich

2-Keto-3-deoxyoctonate ammonium salt

≥97%

Synonym(s):

3-Deoxyoctulosonic acid, KDO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H14O8 · NH3
CAS Number:
Molecular Weight:
255.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

powder

storage temp.

−20°C

SMILES string

N.OCC(O)C1OC(O)(CC(O)C1O)C(O)=O

InChI

1S/C8H14O8.H3N/c9-2-4(11)6-5(12)3(10)1-8(15,16-6)7(13)14;/h3-6,9-12,15H,1-2H2,(H,13,14);1H3

InChI key

UDJBBKGYARWWNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis of KDO Analogues: A significant study focused on the synthesis of analogues of 3-deoxy-D-manno-octulosonic acid (KDO), which includes compounds such as 2-Keto-3-deoxyoctonate ammonium salt. These analogues were evaluated as potential inhibitors of CMP-KDO synthetase, playing a critical role in the development of new therapeutic agents targeting bacterial infections and enhancing understanding of bacterial lipopolysaccharide synthesis (Luthman et al., 1987).

Biochem/physiol Actions

KDO is a sialic acid that is a component of bacterial lipopolysaccharides and a possible target for antibiotic development.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Cipolla et al.
Current drug discovery technologies, 6(1), 19-33 (2009-03-12)
Despite important advances made in the last century, infectious diseases caused by pathogenic microrganisms are still a major threat to human health. This is worsened by the occurrence of new forms of bacterial resistance against antibiotics, that are the main
Cadi Davies et al.
Frontiers in cellular and infection microbiology, 9, 177-177 (2019-06-14)
Campylobacter jejuni outer membrane vesicles (OMVs) contain numerous virulence-associated proteins including the cytolethal distending toxin and three serine proteases. As C. jejuni lacks the classical virulence-associated secretion systems of other enteric pathogens that deliver effectors directly into target cells, OMVs
Chiguang Feng et al.
PloS one, 7(4), e32359-e32359 (2012-04-13)
We previously reported that neuraminidase (NA) pretreatment of human PBMCs markedly increased their cytokine response to lipopolysaccharide (LPS). To study the mechanisms by which this occurs, we transfected HEK293T cells with plasmids encoding TLR4, CD14, and MD2 (three components of
Leticia Martínez-Lapuente et al.
Food research international (Ottawa, Ont.), 96, 235-243 (2017-05-22)
The effect of egg albumin fining, progressive clarification and cross-flow microfiltration on the polysaccharide and proanthocyanidin composition of four red varietal wines was studied in this work. Discriminant analyses were applied to achieve a possible differentiation of the wines according
Blanka Bucsella et al.
Analytical methods : advancing methods and applications, 12(38), 4621-4634 (2020-09-15)
The paper presents a novel instrumental analytical endotoxin quantification assay. It uses common analytical laboratory equipment (HPLC-FLD) and allows quantifying endotoxins (ETs) in different matrices from about 109 EU per mL down to about 40 EU per mL (RSE based).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service