Skip to Content
Merck
All Photos(2)

Key Documents

I3807

Sigma-Aldrich

trans-3-Indoleacrylic acid

98%

Synonym(s):

(2E)-3-(1H-Indol-3-yl)-2-propenoic acid, (E)-3-(1H-Indol-3-yl)-2-propenoic acid, (E)-3-(1H-Indol-3-yl)prop-2-enoic acid, trans-β-Indoleacrylic acid, trans-3-(1H-Indol-3-yl)prop-2-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO2
CAS Number:
Molecular Weight:
187.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

185 °C (dec.) (lit.)

SMILES string

[H]\C(=C(\[H])c1c[nH]c2ccccc12)C(O)=O

InChI

1S/C11H9NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-7,12H,(H,13,14)/b6-5+

InChI key

PLVPPLCLBIEYEA-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Metabolite of tryptophan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Q Marmorstein et al.
The Journal of biological chemistry, 262(10), 4922-4927 (1987-04-05)
We have employed equilibrium dialysis to help study the mechanism by which the unliganded Escherichia coli trp aporepressor is activated by L-tryptophan to the liganded trp repressor. By measuring the relative affinity of L-tryptophan and various tryptophan analogues for the
Huu-Nghi Nguyen et al.
Scientific reports, 9(1), 3166-3166 (2019-03-01)
Matrix-assisted laser desorption/ionisation mass spectrometry imaging (MALDI-MSI) is presently used in physiological evaluations for visualisation of targets in organs. In the present study, MALDI-MSI was used as a visualisation technique to investigate the intestinal absorption of polyphenols. Nifedipine/phytic acid-aided MALDI-MSI
Sergio M Carballo et al.
Food & function, 8(9), 3374-3382 (2017-09-02)
In this work we characterize the interaction of cranberry (Vaccinium macrocarpon) proanthocyanidins (PAC) with bovine serum albumin (BSA) and hen egg-white lysozyme (HEL) and determine the effects of these complexes on macrophage activation and antigen presentation. We isolated PAC from
Chad Robins et al.
Rapid communications in mass spectrometry : RCM, 17(24), 2839-2845 (2003-12-16)
Matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS) with nonpolar matrices has been investigated for its applicability to the characterization of atmospheric resid crude oil fractions. The data obtained by use of nonpolar matrices was compared with that from polar matrices
J K Johnson et al.
Biochemistry, 33(15), 4738-4744 (1994-04-19)
In a previous paper, we demonstrated that the medium-chain fatty acyl-CoA dehydrogenase-catalyzed (MCAD-catalyzed) reductive half-reaction of indolepropionyl-CoA proceeds via formation of a chromophoric intermediary species "X" (absorption maximum = 400 nm) and proposed that the decay of this species might

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service