I3777
Indole-3-acetic hydrazide
97%
Synonym(s):
3-Indolylacetic acid hydrazide, NSC 17812, NSC 63799
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H11N3O
CAS Number:
Molecular Weight:
189.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
142-145 °C (lit.)
SMILES string
NNC(=O)Cc1c[nH]c2ccccc12
InChI
1S/C10H11N3O/c11-13-10(14)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5,11H2,(H,13,14)
InChI key
GYHLCXMCGCVVCG-UHFFFAOYSA-N
Related Categories
Application
Reactant for preparation of:
- Antifungal agents
- Anticonvulsant agents
- Alpha-4 (α4) integrin antagonists
- A fluorescent analogue of UDP-N-acetylglucosamine
- Antioxidant agents with myocardial protection properties
- Antagonist agents of neuropeptide Y receptors
- Antibacterial agents
- Potential monoamine oxidase inhibitors
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service