Skip to Content
Merck
All Photos(1)

Documents

107425

Sigma-Aldrich

Nicotinic hydrazide

97%

Synonym(s):

Nicotinic acid hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
Beilstein:
119299
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

159-161 °C (lit.)

solubility

H2O: soluble 50 mg/mL

SMILES string

NNC(=O)c1cccnc1

InChI

1S/C6H7N3O/c7-9-6(10)5-2-1-3-8-4-5/h1-4H,7H2,(H,9,10)

InChI key

KFUSANSHCADHNJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic hydrazide is a heterocyclic compound that can be used to synthesize Schiff bases.

Application

Nicotinic hydrazide was used in hydrazone library formation. It was used to study the oxidation of isonicotinic acid hydrazide (isoniazid) by horseradish peroxidase.

Biochem/physiol Actions

Nicotinic hydrazide is an inhibitor of peroxidase enzyme. It forms solid metal complexes having strong biological activity.

Preparation Note

Nicotinic hydrazide dissolves in water at a concentration of 50 mg/ml to form a clear, colourless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V Goral et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(4), 1347-1352 (2001-02-15)
Dynamic combinatorial libraries are mixtures of compounds that exist in a dynamic equilibrium and can be driven to compositional self adaptation via selective binding of a specific assembly of certain components to a molecular target. We present here an extension
H A Shoeb et al.
Antimicrobial agents and chemotherapy, 27(3), 399-403 (1985-03-01)
Oxidation of isonicotinic acid hydrazide (isoniazid) by horseradish peroxidase at the expense of H2O2 yielded reactive species which were able to reduce nitroblue tetrazolium and bleach p-nitrosodimethylaniline. Nicotinic acid hydrazide oxidation did not cause these effects. At slightly alkaline pH
Thermo-chemical behavior of solid nicotinic hydrazide metal complexes in correlation with their stoichiometry.
Sekkina MM and El-Azm MG.
Thermochimica Acta, 77(1), 211-218 (1984)
Synthesis, spectroscopic characterization, and crystal structures of Schiff bases derived from nicotinic hydrazide
Diouf F, et al.
IOSR journal of applied chemistry, 1 serie II (2022)
B Toth
Oncology, 38(2), 106-109 (1981-01-01)
Nicotinic acid hydrazide was administered as a 0.125% solution in drinking water continuously for life from 6 weeks of age to randomly bred Swiss albino mice. As a result of treatment, the lung tumor incidence rose from 25 to 76%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service