Skip to Content
Merck
All Photos(3)

Key Documents

P49104

Sigma-Aldrich

Piperonal

99%

Synonym(s):

1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
Beilstein:
131691
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 87 °C)

Quality Level

Assay

99%

form

solid

bp

264 °C (lit.)

mp

35-39 °C (lit.)

SMILES string

[H]C(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2

InChI key

SATCULPHIDQDRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

150.1 °F

Flash Point(C)

65.62 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael S Macias et al.
Forensic science international, 195(1-3), 132-138 (2010-01-02)
Currently, in the field of odor detection, there is generally a wider variation in limit of detections (LODs) for canines than instruments. The study presented in this paper introduces an improved protocol for the creation of controlled odor mimic permeation
Floyd Beckford et al.
Journal of inorganic biochemistry, 105(8), 1019-1029 (2011-06-15)
The synthesis and characterization of a number of organometallic ruthenium(II) complexes containing a series of bidentate thiosemicarbazone ligands derived from piperonal is reported. The structure of compounds have been confirmed by spectroscopic analysis (IR and NMR) as well as X-ray
S B Stanfill et al.
Journal of agricultural and food chemistry, 48(4), 1298-1306 (2000-04-25)
Little is known about the possible health effects associated with inhaling alkenylbenzenes through cigarette smoking, even though these flavor-related compounds have known toxic effects in animals. We developed a rapid and sensitive solid-phase extraction (SPE) method to quantify seven alkenylbenzenes
K C Nicolaou et al.
Journal of the American Chemical Society, 130(39), 13110-13119 (2008-09-06)
The molecular design, chemical synthesis, and biological evaluation of two distinct series of platensimycin analogues with varying degrees of complexity are described. The first series of compounds probes the biological importance of the benzoic acid subunit of the molecule, while
H Hatt et al.
Cellular and molecular biology (Noisy-le-Grand, France), 45(3), 285-291 (1999-07-01)
The human olfactory system can recognize and discriminate a large number of different odorant molecules. The detection of chemically distinct odorants begins with the binding of an odorant ligand to a specific receptor protein on the olfactory neuron cell surface.

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service