I1000
Indole-3-acetaldehyde–sodium bisulfite addition compound
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C10H9NO · NaHSO3
CAS Number:
Molecular Weight:
263.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
impurities
≤15% excess sodium bisulfite
Quality Level
storage temp.
2-8°C
SMILES string
[Na].OC(Cc1c[nH]c2ccccc12)S(O)(=O)=O
InChI
1S/C10H11NO4S.Na/c12-10(16(13,14)15)5-7-6-11-9-4-2-1-3-8(7)9;/h1-4,6,10-12H,5H2,(H,13,14,15);/q;+1/p-1
InChI key
WYKPCRCESPDDQX-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Reactant for preparation of potential neurotransmitter analogs
- Reactant for Strecker reaction with cyanide-11C
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hayes, R. L.; et al.
The International Journal of Applied Radiation and Isotopes, 29, 186-186 (1978)
Schlecht, M., F.; Giandinoto, S.
Heterocycles, 25, 485-485 (1987)
Davide Marangon et al.
Glia, 68(10), 2001-2014 (2020-03-13)
In the last decade, microRNAs have been increasingly recognized as key modulators of glial development. Recently, we identified miR-125a-3p as a new player in oligodendrocyte physiology, regulating in vitro differentiation of oligodendrocyte precursor cells (OPCs). Here, we show that miR-125a-3p
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service