GF73794741
Gold
tube, 200mm, outside diameter 2.0mm, inside diameter 1.8mm, wall thickness 0.1mm, as drawn, 99.95%
Synonym(s):
Gold, AU007200
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
99.95%
form
tubes
manufacturer/tradename
Goodfellow 737-947-41
resistivity
2.05 μΩ-cm, 0°C
L × wall thickness
200 mm × 0.1 mm
O.D. × I.D.
2.0 mm × 1.8 mm
bp
2808 °C (lit.)
mp
1063 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[Au]
InChI
1S/Au
InChI key
PCHJSUWPFVWCPO-UHFFFAOYSA-N
Related Categories
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of biomedical nanotechnology, 10(6), 982-992 (2014-04-23)
Electroporation figured prominently as an effective nonviral gene delivery approach for its balance on the transfection efficiency and cell viability, no restrictions of probe or cell type, and operation simplicity. The commercial electroporation systems have been widely adopted in the
Journal of nanoscience and nanotechnology, 14(7), 4906-4910 (2014-04-25)
Effect of structure and size of Au nanoparticles (AuNPs) on fluorescence behavior of uranine was examined. Confeito-like AuNPs with different sizes (30 nm, 60 nm and 100 nm, respectively) had plasmon absorption bands at 555, 600 and 660 nm, while
Journal of nanoscience and nanotechnology, 14(7), 5151-5156 (2014-04-25)
An immunochromatographic test strip using gold nanoparticles-staphylococcus aureus monoclonal antibody conjugates was developed for the rapid and convenient detection of staphylococcus aureus based on a double-antibody sandwich format. The detection limit and the detection rate of this test strip is
Journal of nanoscience and nanotechnology, 14(7), 4813-4819 (2014-04-25)
The applications of nanoparticles (NPs) for improved therapeutics are at the forefront of cancer nanotechnology. Gold nanoparticles (GNPs) have been extensively used due to their ability to act as both an anticancer drug carrier in chemotherapy and as a dose
Journal of nanoscience and nanotechnology, 14(6), 4124-4138 (2014-04-18)
Gold nanostructures with promising applications in biomedical field have attracted great attention. However, some fundamental questions other than the development of novel applications should be elucidated before they can actually serve as biomedicines in the clinic. Bio-safety is one of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service