Skip to Content
Merck
All Photos(4)

Key Documents

379646

Sigma-Aldrich

Fullerene-C60

greener alternative

99.5%

Synonym(s):

Buckminsterfullerene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60
CAS Number:
Molecular Weight:
720.64
Beilstein:
5901022
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

99.5%

form

crystalline powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>280 °C (lit.)

solubility

organic solvents: soluble
organic solvents: soluble

Orbital energy

HOMO 6.1 eV 
LUMO 4.5 eV 

OLED Device Performance

ITO/CuPc/NPD/Alq3/C60/Mg:Ag

  • Color: green
  • Max. Luminance: 17200 Cd/m2

ITO/NPD/Alq3/C60/LiF:Al:Ag
  • Color: green
  • Max. Luminance: 7000 Cd/m2
  • Turn-On Voltage: <5 V

ITO/TPD/Alq3/C60/LiF/Al/Ag
  • Color: green
  • Max. Luminance: 4000 Cd/m2
  • Turn-On Voltage: <5 V

greener alternative category

SMILES string

c12c3c4c5c1c6c7c8c2c9c%10c3c%11c%12c4c%13c%14c5c%15c6c%16c7c%17c%18c8c9c%19c%20c%10c%11c%21c%22c%12c%13c%23c%24c%14c%15c%25c%16c%26c%17c%27c%18c%19c%28c%20c%21c%29c%22c%23c%30c%24c%25c%26c%31c%27c%28c%29c%30%31

InChI

1S/C60/c1-2-5-6-3(1)8-12-10-4(1)9-11-7(2)17-21-13(5)23-24-14(6)22-18(8)28-20(12)30-26-16(10)15(9)25-29-19(11)27(17)37-41-31(21)33(23)43-44-34(24)32(22)42-38(28)48-40(30)46-36(26)35(25)45-39(29)47(37)55-49(41)51(43)57-52(44)50(42)56(48)59-54(46)53(45)58(55)60(57)59

InChI key

XMWRBQBLMFGWIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fullerene-C60 (C60, 99.5%) belongs to the class of carbonaceous materials. It is approximately 0.7 nm in diameter. It is a hollow, icosahedrally-shaped, closed-cage structure consisting of 60 sp2 hybridized carbon atoms which can be utilized for the preparation of novel carbon nanomaterials.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

C60 has a broad range of applications which include catalysis, optical materials, polymer modification, solar cells, fuel cells, lubricants, cosmetics, and biomedical applications.
It can be used for a variety of alternative energy applications, which include lithium-ion batteries, organic photovoltaic cells (OPVs), heterojunction solar cells, polymeric photodetectors, photodiodes, and photocatalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced infra-red emission from sub-millimeter microelectromechanical systems micro hotplates via inkjet deposited carbon nanoparticles and fullerenes.
Luca AD, et al.
Journal of Applied Physics, 113 null
Application of fullerene-added nano-oil for lubrication enhancement in friction surfaces.
Lee J, et al.
Tribology International, 42(3), 440-447 (2009)
Polymer-fullerene composite solar cells.
Thompson BC and Frechet JMJ
Angewandte Chemie (International ed. in English), 47(1), 58-77 (2008)
Application of nanotechnology in cosmetics.
Mu Li and Sprando RL
Pharmaceutical Research, 27(8), 1746-1749 (2010)
Double-cable polymers for fullerene based organic optoelectronic applications
Cravino A and Sariciftci NS
Journal of Materials Chemistry, 12(7), 1931-1943 (2002)

Articles

Carbon nanotubes are materials that possess remarkable properties and offer extraordinary possibilities.

Find various photovoltaic and bioscience-based applications of fullerenes.

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

Professor Aran discusses engineering graphene-based materials through careful functionalization, enabling diverse applications.

See All

Related Content

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Batteries, fuel cells, and supercapacitors rely on electrochemical energy production. Understand their operation and electron/ion transport separation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service