Skip to Content
Merck
All Photos(2)

Key Documents

GF38495885

Manganese

rod, 50mm, diameter 4.0mm, cast, 99.5%

Synonym(s):

Manganese, MN007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.5%

form

rod

manufacturer/tradename

Goodfellow 384-958-85

resistivity

185 μΩ-cm, 20°C

L × diam.

50 mm × 4.0 mm

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monika Meyer-Baron et al.
Neurotoxicology, 36, 1-9 (2013-02-20)
Results from a meta-analysis of aggregated data provoked a new analysis using individual data on the neuropsychological performance of occupationally exposed workers. Data from eight studies examining 579 exposed and 433 reference participants were included, 28 performance variables analyzed. The
Håkan Nilsson et al.
Biochimica et biophysica acta, 1837(8), 1257-1262 (2014-04-15)
The binding affinity of the two substrate-water molecules to the water-oxidizing Mn₄CaO₅ catalyst in photosystem II core complexes of the extremophilic red alga Cyanidioschyzon merolae was studied in the S₂ and S₃ states by the exchange of bound ¹⁶O-substrate against
Mohammad Mahdi Najafpour et al.
Photosynthesis research, 114(1), 1-13 (2012-09-04)
The resolution of Photosystem II (PS II) crystals has been improved using isolated PS II from the thermophilic cyanobacterium Thermosynechococcus vulcanus. The new 1.9 Å resolution data have provided detailed information on the structure of the water-oxidizing complex (Umena et
Karin Tuschl et al.
International review of neurobiology, 110, 277-312 (2013-11-12)
Manganese (Mn) is an essential trace metal that is pivotal for normal cell function and metabolism. Its homeostasis is tightly regulated; however, the mechanisms of Mn homeostasis are poorly characterized. While a number of proteins such as the divalent metal
Mirna Lechpammer et al.
Annals of neurology, 75(4), 608-612 (2014-03-07)
We followed a patient with manganese transporter deficiency due to homozygous SLC30A10 mutations from age 14 years until his death at age 38 years and present the first postmortem findings of this disorder. The basal ganglia showed neuronal loss, rhodanine-positive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service