Skip to Content
Merck
All Photos(2)

Key Documents

GF11021094

Manganese

rod, 10mm, diameter 4.0mm, cast, 99.5%

Synonym(s):

Manganese, MN007910

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

99.5%

form

rod

manufacturer/tradename

Goodfellow 110-210-94

resistivity

185 μΩ-cm, 20°C

L × diam.

10 mm × 4.0 mm

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ebany J Martinez-Finley et al.
Free radical biology & medicine, 62, 65-75 (2013-02-12)
Manganese (Mn) is an essential dietary nutrient, but an excess or accumulation can be toxic. Disease states, such as manganism, are associated with overexposure or accumulation of Mn and are due to the production of reactive oxygen species, free radicals
Mirna Lechpammer et al.
Annals of neurology, 75(4), 608-612 (2014-03-07)
We followed a patient with manganese transporter deficiency due to homozygous SLC30A10 mutations from age 14 years until his death at age 38 years and present the first postmortem findings of this disorder. The basal ganglia showed neuronal loss, rhodanine-positive
Srinivasan V Narayanan et al.
Stroke, 46(6), 1626-1632 (2015-04-25)
Nuclear erythroid 2 related factor 2 (Nrf2) is an astrocyte-enriched transcription factor that has previously been shown to upregulate cellular antioxidant systems in response to ischemia. Although resveratrol preconditioning (RPC) has emerged as a potential neuroprotective therapy, the involvement of
Monika Meyer-Baron et al.
Neurotoxicology, 36, 1-9 (2013-02-20)
Results from a meta-analysis of aggregated data provoked a new analysis using individual data on the neuropsychological performance of occupationally exposed workers. Data from eight studies examining 579 exposed and 433 reference participants were included, 28 performance variables analyzed. The
Håkan Nilsson et al.
Biochimica et biophysica acta, 1837(8), 1257-1262 (2014-04-15)
The binding affinity of the two substrate-water molecules to the water-oxidizing Mn₄CaO₅ catalyst in photosystem II core complexes of the extremophilic red alga Cyanidioschyzon merolae was studied in the S₂ and S₃ states by the exchange of bound ¹⁶O-substrate against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service