Skip to Content
Merck
All Photos(2)

Documents

GF31110859

Carbon

foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.997%

Synonym(s):

Activated charcoal, C 000411, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

99.997%

form

foil

autoignition temp.

842 °F

manufacturer/tradename

Goodfellow 311-108-59

resistivity

1375 μΩ-cm, 20°C (graphite)

size × thickness

100 × 100 mm × 0.25 mm

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Investigation of delta -electron emission in collisions of highly charged fast Ne projectiles with carbon-foil targets.
Schiwietz et al.
Physical review. B, Condensed matter, 41(10), 6262-6271 (1990-04-01)
I Dietrich et al.
Ultramicroscopy, 3(2), 185-189 (1978-01-01)
Radiation damage on a holey carbon foil was investigated in an electron microscope with a superconducting lens system, where the temperature of the specimen and its environment initially was 4 K. Due to an electron dose of 2 X 10(4)
Doubly differential secondary-electron yields following 8-MeV/u U68+- and 3.5-MeV/u U38+-ion impact on a thin carbon-foil target.
Schneider et al.
Physical review. A, Atomic, molecular, and optical physics, 47(5), 3945-3950 (1993-05-01)
M C Ledbetter et al.
Proceedings of the National Academy of Sciences of the United States of America, 84(1), 85-88 (1987-01-01)
Cluster ions containing 50, 100, or 200 water molecules and one proton are accelerated to over 275 kV and impacted on thin carbon films on which tobacco mosaic virus has been dispersed. After bombardment the films are examined with a
Z oscillation of mean charges of energetic ions emerging from a carbon foil: Correlation with the shell structure of ions.
Shima et al.
Physical review. A, General physics, 39(8), 4316-4318 (1989-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service