![Carbon rod, 100mm, diameter 25mm, graphite, 99.95%](/deepweb/assets/sigmaaldrich/product/structures/214/104/6859671a-6bb1-41d7-a654-482af53dd632/640/6859671a-6bb1-41d7-a654-482af53dd632.png)
Carbon
![Carbon rod, 100mm, diameter 25mm, graphite, 99.95%](/deepweb/assets/sigmaaldrich/product/structures/214/104/6859671a-6bb1-41d7-a654-482af53dd632/640/6859671a-6bb1-41d7-a654-482af53dd632.png)
Synonym(s):
Carbon, Activated charcoal, Charcoal activated
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Element
Quality Segment
Showing 1-20 of 142
Page 1 of 8
Compare
Product Number
Description
Pricing
Page 1 of 8