Skip to Content
Merck
All Photos(1)

Key Documents

D70406

Sigma-Aldrich

3,4-Dichlorophenol

99%

Synonym(s):

3,4-DCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
1907693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

~253 °C (lit.)

mp

65-67 °C (lit.)

SMILES string

Oc1ccc(Cl)c(Cl)c1

InChI

1S/C6H4Cl2O/c7-5-2-1-4(9)3-6(5)8/h1-3,9H

InChI key

WDNBURPWRNALGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,4-Dichlorophenol (3,4-DCP) is used in the synthesis of (Z)-5-arylmethylidene rhodanines as anti-methicillin-resistant Staphylococcus aureus (MRSA) agent. It can also be used in the preparation of 2-(3,4-dichlorophenoxy)-N-(2-morpholin-4-ylethyl)acetamide for the treatment of inflammatory pain.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis and SAR study of phenylalanine-derived (Z)-5-arylmethylidene rhodanines as anti-methicillin-resistant Staphylococcus aureus (MRSA) compounds.
Patel BA, et al.
Bioorganic & Medicinal Chemistry Letters, 23(20), 5523-5527 (2013)
Discovery of 2-(3, 4-dichlorophenoxy)-N-(2-morpholin-4-ylethyl) acetamide: A selective ?1 receptor ligand with antinociceptive effect.
Navarrete-Vazquez G, et al.
Biomedicine and Pharmacotherapy, 79(20), 284-293 (2016)
Julien Michel et al.
Journal of the American Chemical Society, 131(18), 6356-6357 (2009-05-07)
There is great interest in molecules capable of inhibiting the interactions between p53 and its negative regulators hDM2 and hDMX, as these molecules have validated potential against cancers in which one or both oncoproteins are overexpressed. We reported previously that
F O Bryant et al.
Applied and environmental microbiology, 57(8), 2293-2301 (1991-08-01)
The reductive dechlorination of pentachlorophenol (PCP) was investigated in anaerobic sediments that contained nonadapted or 2,4- or 3,4-dichlorophenol (DCP)-adapted microbial communities. Adaptation of sediment communities increased the rate of conversion of 2,4- or 3,4-DCP to monochlorophenols (CPs) and eliminated the
J H Carey et al.
Canadian journal of physiology and pharmacology, 62(8), 971-975 (1984-08-01)
The disappearance rates of 2,4- and 3,4-dichlorophenol in a small stream were studied and were shown to be first order with respect to either distance or time of flow. Both chlorophenols disappeared at approximately the same rate with average half-lives

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service