Skip to Content
Merck
All Photos(2)

Key Documents

D35201

Sigma-Aldrich

N,N′-Dibenzylethylenediamine

97%

Synonym(s):

DBED

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NHCH2CH2NHCH2C6H5
CAS Number:
Molecular Weight:
240.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.565 (lit.)

bp

195 °C/4 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

C(CNCc1ccccc1)NCc2ccccc2

InChI

1S/C16H20N2/c1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16/h1-10,17-18H,11-14H2

InChI key

JUHORIMYRDESRB-UHFFFAOYSA-N

General description

N,N′-Dibenzylethylenediamine, also known as DBED, is a donor ligand often used in the synthesis of coordination compounds and in the formation of stable metal complexes such as nickel (II) carboxylate complex.

Application


  • Ring-Opening Polymerization of rac-Lactide Catalyzed by Octahedral Nickel Carboxylate Complexes: Focuses on a complex involving N,N′-dibenzylethylenediamine as a syndioselective catalyst in polymer synthesis (A Nikiforov et al., 2023).

  • Synthesis, Crystal Structures and Catalytic Activities of Two Copper Coordination Compounds Bearing an N, N′-Dibenzylethylenediamine Ligand: Examines copper coordination compounds that incorporate N,N′-dibenzylethylenediamine and their structural and catalytic properties (C Liu et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzathine interference in the EMIT-st urine amphetamine assay.
N R Badcock et al.
Clinical chemistry, 33(6), 1080-1080 (1987-06-01)
S Meier et al.
Journal of animal science and biotechnology, 11, 17-17 (2020-02-12)
In seasonal, pasture-based, dairy production systems, cows must recover from calving and become pregnant within two to 3 months. To achieve this, the uterus must involute and ovulation must occur and continue at regular intervals. As these processes are affected
Renata Pereira Lopes et al.
Talanta, 89, 201-208 (2012-01-31)
A multiclass method has been optimized and validated for the simultaneous determination of 20 veterinary drug residues belonging to several classes, as quinolones, sulfonamides, macrolides, anthelmintics, avermectins and diamino derivatives, and benzathine, used as a marker of the presence of
Stephen Nurse-Findlay et al.
PLoS medicine, 14(12), e1002473-e1002473 (2017-12-28)
Benzathine penicillin G (BPG) is the only recommended treatment to prevent mother-to-child transmission of syphilis. Due to recent reports of country-level shortages of BPG, an evaluation was undertaken to quantify countries that have experienced shortages in the past 2 years
G S Bassi et al.
Progress in neuro-psychopharmacology & biological psychiatry, 77, 146-154 (2017-04-10)
The production of unconditioned defensive behaviors has been related to the amygdala, a key component of the encephalic aversion system. Microinjection of the neuropeptide substance P (SP) in the amygdala elicits defensive behaviors via the activation of type 1 neurokinin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service